Search Thermo Fisher Scientific
Optimize your experiments to get the best results. We’ve compiled a detailed knowledgebase of the top tips and tricks to meet your research needs.
View the relevant questions below:
Having problems with your experiment? Visit our
Recommended Maintenance Schedule | |
---|---|
Power on/off the computer controlling the instrument | Weekly |
Check computer disk space. If necessary, archive or back up your experiment files and instrument settings | Weekly |
Background calibration | Every month |
Run disk cleanup and disk defragmentation | Every month |
Pure dye calibrations | Every 6 months |
ROI calibration | Every 6 months |
Uniformity calibration | Every 6 months |
Normalization calibration | Every 6 months |
RNaseP instrument verification | After the instrument has been moved, or as needed to verify instrument performance |
The uniformity calibration generates data that allow the software to compensate for the physical effects of the ViiA™ 7 system filters using the ROI plate. The normalization calibration generates factors that the software uses when comparing data from multiple ViiA™ 7 instruments within a study. Two normalization plates are provided in the calibration kit, one containing FAM™/ROX™ dyes and the other with VIC®/ROX™ dyes.
The calibration plates can be stored and reused three times for up to 6 months after you first open them, so make sure to return them to their original packaging and return them to –20°C storage until the next use. (Note: Due to the small volumes, calibration array cards cannot be reused. Please discard them after use.) If needed, you can make your own background plate using deionized water. Please follow the directions in the ViiA™ 7 Real-Time PCR System Installation, Networking, and Maintenance Guide for more details.
Please ask your dye manufacturer for recommendations. If there is no specific recommendation available, try TE Buffer (pH 8.0) to start.
The RNaseP verification plate contains template, master mix, and a TaqMan® assay for RNaseP. It is used to verify that the instrument is performing to specifications. If you have reason to suspect there is something wrong with the instrument, or if you want to rule out a chemistry issue, the RNaseP plate is a good way to test the system. The RNaseP verification plate is a single-use plate.
You can create your own background calibration plate using the directions in the ViiA™ 7 Real-Time PCR System Maintenance Guide (Appendix C). For all other calibration plates, you can purchase a calibration kit, or the individual plates, as required.
In a touchdown PCR experiment, you will change either the temperature or the time of a particular PCR step with every cycle. Most commonly, the annealing temperature is adjusted throughout the experiment, such that the specificity is higher in the early cycles and the efficiency in the later cycles. In this example, we will set the method to do the following:
40 cycles ![]() |
|
You will need an oligo sequence with the custom dye but without the quencher molecule. For each custom dye, prepare samples in a concentration range of 100–2,000 nM, choosing a 2–3-fold difference in dilution points. Use either the background buffer provided in the calibration kit or your own buffer to dilute the oligo. A full plate is not needed; see the example below.
In this example, sample concentrations would be 100, 200, 400, 800, and 1,600 (20 µL/well). Note: The volume would be the same for a 96-well or 384-well plate.
The ViiA™ 7 Real-Time PCR System can store up to 100 runs on the instrument itself.
The following volumes are supported for each instrument block:
Yes. The ViiA™ 7 system can collect data at multiple steps in the amplification stage. You will need to turn on the data collection for every step of interest. In the analysis, only one set of data can be displayed at a time. To change the data set used for analysis, go to Analysis Settings → Ct Settings and change the dropdown under ‘Data Step Selection’. In this example, data collection is turned on at both 63°C and 72°C.
MicroAmp 96-Well Tray for VeriFlex Blocks (Cat. No. 4379983) is needed to load 1 or 2 MicroAmp Optical 8-Tube Strips with Attached Domed Caps on the ProFlex, SimpliAmp, and Veriti thermal cycler 96-well blocks.
Yes. The tubes are DNase/RNase/Human DNA free, as they are manufactured in a Class 100K ISO certified clean room UK production facility.
The MicroAmp 8-Tube Strip with Attached Domed Caps is compatible with the following instruments:
- ProFlex 3 x 32-well PCR System
- ProFlex 96-well
- Veriti 96-well Thermal Cycler
- SimpliAmp Thermal Cycler
- MiniAmp Thermal Cycler
- MiniAmp Plus Thermal Cycler
- 2720 Thermal Cycler
- GeneAmp PCR System 9700, 96-well
These new strips have new features including a graduated 20 µL measuring mark for visual verification and dual end tabs for better labeling and handling. In addition, the individual caps help prevent cross contamination and reduce sample evaporation.
Please refer to the table on Page 2 of this flyer for compatibility information.
Yes. The tubes are DNase/RNase/Human DNA free, as they are manufactured in a Class 100K ISO certified clean room UK production facility.
These new strips have new features including a graduated 20 μL measuring mark for visual verification and dual end tabs for better labeling and handling. In addition, the individual caps help prevent cross contamination and reduce sample evaporation.
MicroAmp 96-Well Tray for VeriFlex Blocks (Cat. No. 4379983) is needed to load 1 or 2 MicroAmp Optical 8-Tube Strips with Attached Optical Caps on the ProFlex, SimpliAmp, MiniAmp, MiniAmp Plus, and Veriti Thermal Cycler 96-well blocks, and QuantStudio 3 and QuantStudio 5 blocks.
The MicroAmp Optical 8-Tube Strip with Attached Optical Caps is compatible with the following instruments:
- ProFlex 3 x 32-well PCR System
- ProFlex 96-well PCR System
- Veriti 96-well Thermal Cycler
- SimpliAmp Thermal Cycler
- MiniAmp Thermal Cycler
- MiniAmp Plus Thermal Cycler
- 2720 Thermal Cycler
- GeneAmp PCR System 9700, 96-well
- 7000 PCR System
- 7300 Real-Time PCR System
- 7500 Real-Time PCR system
- QuantStudio Real-Time PCR Systems
- ViiA7 Real-Time PCR System, 96-well
Yes. The tubes are DNase/RNase/Human DNA free, as they are manufactured in a Class 100K ISO certified clean room UK production facility.
These new strips have new features including a graduated 20 µL measuring mark for visual verification and dual end tabs for better labeling and handling. In addition, the individual caps help prevent cross contamination and reduce sample evaporation.
Please refer to the table on Page 2 of this flyer for compatibility information.
Yes. The tubes are DNase/RNase/Human DNA free, as they are manufactured in a Class 100K ISO certified clean room UK production facility.
These new strips have new features including a graduated 20 µL measuring mark for visual verification and dual end tabs for better labeling and handling. In addition, the individual caps help prevent cross contamination and reduce sample evaporation.
For genotyping data, we recommend TaqMan® Genotyper Software. For relative quantitation, we recommend ExpressionSuite™ Software.
For Research Use Only. Not for use in diagnostic procedures.