c7c8 - Key/Specs/Resources/Appl/Tech/Doc/Contact
With time-to-yield being critical to profitability, it is too costly for advanced foundries, integrated device manufacturers and fabless companies to wait until after dicing and packaging to identify the source of electrical faults. The Thermo Scientific Meridian WS-DP system enables faster defect localization by using production testers, load boards, and probe cards.
All diagnostic options from the Meridian product line are available for the Meridian WS-DP, including: LVx, emission, LADA, and OBIRCH.
The Meridian WS-DP system provides:
  • Ability to identify systematic defects
  • Significantly increased electrical  measurement throughput
  • Improved defect localization success rate
  • Reduced physical analysis time
Capabilities:
  • Standard InGaAs-based emission detection or
  • High sensitivity extended-wavelength DBX-based emission detection
  • Patented, industry-proven "Point & Click" Solid Immersion Lens (SIL)
  • Rapid die-to-die indexing
  • Dynamic fault coverage with high-speed ATE interface
  • Laser scanning microscope (LSM) for static and dynamic analysis
  • Compatibility with most popular third-party EDA applications
Key Features
Laser voltage imaging
Laser voltage imaging (LVI), shows the physical locations of transistors that are active at a specific frequency on the Meridian WS-DP system.
CW laser voltage probing
Continuous wave laser voltage probing (CW-LVP) acquires functional waveform data on the Meridian WS-DP system.
Photon emission microscopy
Photon emission microscopy (PEM) on the Meridian platform is based on an optimized combination of a high sensitivity InGaAs or DBX camera and high numerical aperture (NA) aberration-corrected optics. The Meridian WS-DP system provides high-resolution through-silicon imaging for backside device analysis. Low noise and high sensitivity enable emission data with unmatched signal-to-noise ratios, resulting in rapid, transistor-level fault detection. The DBX configuration provides industry-leading results even on sub-0.5 Vdd devices.
Specifications
Style Sheet for Products Table Specifications
MechanicalATE compatibility
Commercially available ATE testers & customized solutions
Probe Card / Load Board
  • 9.5", 12", 18" round probe cards
  • Direct probe load boards
  • 440mm & 300mm probe cards
  • Customized solutions available for non-standard geometries
Probe pin count
Probe arrays of <100 to >10,000 pins
Wafer Stage vacuum
20lbs vacuum force for wafer stability
Motion ControlDie-to-die stepping
Supported and integrated into Sierra software
Platen motion
  • +/- 5 degrees of rotation
  • 16um theta resolution
Wafer / DUT supported
  • 9.5", 12", 18" round probe cards
  • Direct probe load boards
  • 440mm & 300mm probe cards
  • Customized solutions available for non-standard geometries
XYZ microscope stage accuracy
1 um
XY wafer stage accuracy
5um
Die size compatibility
A wide range of solutions
Style Sheet for Komodo Tabs
Resources
Applications
Semiconductor-RD_1920x1080
Semiconductor research and development
Innovation starts with research and development. Learn more about solutions to help you understand innovative structures and materials at the atomic level.
Semiconductor Failure Analysis
Semiconductor Failure Analysis
Complex semiconductor device structures result in more places for defects to hide. Learn more about failure analysis solutions to isolate, analyze, and repair defects.
Techniques
Optical Fault Isolation
Increasingly complex designs complicate fault and defect isolation in semiconductor manufacturing. Optical fault isolation techniques allow you to analyze the performance of electrically active devices to locate critical defects that cause device failure.
Learn more ›
Optical Fault Isolation
Increasingly complex designs complicate fault and defect isolation in semiconductor manufacturing. Optical fault isolation techniques allow you to analyze the performance of electrically active devices to locate critical defects that cause device failure.
Learn more ›
Style Sheet to change H2 style to p with em-h2-header class
Contact us
Electron microscopy services for
semiconductors
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
Learn more ›
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards