Organism TypePrimate - Non HumanA COS-like Monkey kidney cell line, CMT3, was transfected by the standard CaPO4 method with plasmids pCMVgagpolrre-r and pCMVrev and a selective marker plasmid that expresses the hygromycin resistance gene to generate cell line B4.14. The pCMVrev gene is required for efficient expression of the gag gene. The cells were maintained in Iscove’s modified Dulbecco’s medium with 10% fetal calf serum, 50 mg of gentamicin per ml, and 200 mg of hygromycin B per ml.
TissuesKidney
PhenotypeAdherent
Primaryyes
ApplicationTransient Transfection
Products
Catalog # Name Size Price (USD) Qty
18292011 Lipofectin™ Transfection Reagent, 1 mL Each
678.00
Every step of the way, a wide range of cell health products
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.