Alternaria alternata Allergen Facts, Symptoms, and Treatment
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Access videos and webinars delivered by key experts in the field of allergy.
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Access videos and webinars delivered by key experts in the field of allergy.
The most prevalent mold in dry, warm climates, Alternaria alternata has air spores that peak in the afternoon and typically disseminate in warm, dry air.1,2 Therefore, in temperate climates, Alternaria alternata spore counts are usually highest in the summer.2 Those allergic to this mold may experience symptoms after inhaling its spores.3 Alternaria alternata is mainly an outdoor fungus that typically grows on vegetation.4 However, the species can also be found indoors, where it prefers humid locations such as bathrooms and often produces large brown spores that are a well-known cause of allergy and asthma.4,5 That said, indoor Alternaria alternata concentrations are typically impacted by the current amount of outdoor concentrations. That is, if there are considerable spores in the outdoor air, indoor levels may be high as well.4
Alternaria thrives at temps of 20 to 25 C (68 to 77 F), but it can survive temperatures between 1 and 35 C (roughly 34 and 95 F).6 Mold sensitization rates vary according to different studies, but allergy to Alternaria alternata seems to affect roughly 5 percent of people and is strongly associated with allergic rhinitis (aka hay fever) and asthma.7 In children, the prevalence of reported sensitization varies dramatically, from less than 1 percent in Austria to 50 percent in the U.S. state of Arizona. And according to a large study of children with asthma living in inner cities in the United States, Alternaria was the most common mold allergy, with 38 percent of study participants incurring a positive skin test to this allergen.2 Alternaria allergy paired with Alternaria exposure is a risk factor for the development and worsening of allergic diseases such as allergic rhinitis and asthma, and it can cause severe asthma exacerbations.4
Alternaria is often found in soil and on mediums such as plants, cereal grains, grass, corn silage, rotten wood, bricks, canvas, iron, compost, and bird nests.2,6 Plus, Alternaria is a plant pathogen and can infect more than 4,000 plant species, causing significant damage to grains, fruits, and vegetables, making it responsible for 20 percent of agricultural yield losses.6 In fact, Alternaria molds grow on a host of foods such as tomatoes, cucumbers, cauliflowers, peppers, apples, melons, tangerines, oranges, lemons, and sunflower seeds.13 That said, mold reproduces via spores, which can be transported by air, water, and insects.8,14 So even if a fungus originates outdoors, it often can enter a dwelling through a variety of means, including doorways, windows, vents, and heating and air conditioning systems.15 Therefore, Alternaria alternata can be found indoors on surfaces such as drywall, ceiling tiles, wood, carpet, wallpaper, textiles, window frames, and materials within heating and air conditioning systems.5,6
If you are allergic to molds, your healthcare provider may recommend a plan that includes the following.3,8,16,17,18
Your healthcare provider may direct you to employ one of the following therapies to improve your allergy symptoms:
Mold allergy symptoms can range from mild to severe and vary person to person.3 Reactions can happen almost immediately after exposure, or they can be delayed. Symptoms are most common in mid-summer to early fall, but since molds grow both indoors and out, allergic reactions can occur all year.8
Symptoms typically include one or more of the following:3,9
Mold sensitization is also a major risk factor for developing upper and lower respiratory diseases such as asthma.2 In fact, exposure to Alternaria mold is related to potentially life-threatening asthma.4 Other reactions to Alternaria can include allergic rhinitis (aka hay fever), allergic sinusitis, and fungal ball production in the sinuses.10
A small percentage of asthmatics with inhaled mold allergy can also develop allergic urticaria (aka hives) when they eat or drink anything containing yeast or mold.11 In addition to causing allergic reactions, molds can also lead to infections along with toxic reactions.12
Together with your symptom history, skin-prick testing or specific IgE blood testing can help determine if you are allergic to a particular allergen. If you are diagnosed with an allergy, your healthcare provider will work with you to create a management plan.
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
Some people with an Alternaria alternata allergy may also experience symptoms when exposed to other types of mold. This is called cross-reactivity and occurs when your body's immune system identifies the proteins, or components, in different substances as being structurally similar or biologically related, thus triggering a response.4
Knowing the proteins, or components, within each allergen that are triggering your symptoms can help guide your management plan. With that in mind, and based on your symptom history, your healthcare provider may suggest something called a specific IgE component test, which can help reveal your potential risk profile.4 Results from this test can also help your healthcare provider decide if allergen immunotherapy may reduce your symptoms.16
Already have your specific IgE component test results?
Your component test results will include the name of the components (a series of letters and numbers). Your healthcare provider will likely review the results with you, but here you'll find an at-a-glance breakdown you can use as a reference for the Alt a 1 component:4,16
rAlt a 1
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.