Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
This affinity purified antibody is directed against human AP3D1. A BLAST analysis was used to suggest reactivity with this protein from human, chimpanzee and chicken based on 100% homology for the immunogen sequence. Cross reactivity with AP3D1 protein from mouse, rat, dog and cow may occur as sequence homology varies by only one amino acid residue in this sequence as indicated by BLAST analysis. Reactivity with AP3D1 proteins from other sources is not known.
AP3D1 is a subunit of the AP3 adaptor-like complex, which is not associated with clathrin. The AP3D1 subunit is implicated in intracellular biogenesis and trafficking of pigment granules and possibly platelet dense granules and neurotransmitter vesicles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adapter-related protein complex 3 subunit delta-1; adaptin, delta; adaptor related protein complex 3 subunit delta 1; adaptor related protein complex 3, delta 1 subunit; Adaptor-related protein complex 3 subunit delta-1; adaptor-related protein complex 3, delta 1 subunit; AP-3 complex delta subunit, partial CDS; AP-3 complex subunit delta; AP-3 complex subunit delta-1; ap3; delta adaptin; Delta-adaptin; delta-adaptin, partial CDS; subunit of putative vesicle coat adaptor complex AP-3
Gene Aliases: ADTD; AP3D1; hBLVR; PRO0039
UniProt ID: (Human) O14617
Entrez Gene ID: (Human) 8943, (Chicken) 420079
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support