Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MISPVLILFS SFLCHVAIAG RTCPKPDDLP FSTVVPLKTF YEPGEEITYS CKPGYVSRGG MRKFICPLTG LWPINTLKCT PRVCPFAGIL ENGAVRYTTF EYPNTISFSC NTGFYLNGAD SAKCTEEGKW SPELPVCAPI ICPPPSIPTF ATLRVYKPSA GNNSLYRDTA VFECLPQHAM FGNDTITCTT HGNWTKLPEC REVKCPFPSR PDNGFVNYPA KPTLYYKDKA TFGCHDGYSL DGPEEIECTK LGNWSAMPSC KASCKVPVKK ATVVYQGERV KIQEKFKNGM LHGDKVSFFC KNKEKKCSYT EDAQCIDGTI EVPKCFKEHS SLAFWKTDAS DVKPC
Apolipoprotein H has been implicated in a variety of physiologic pathways including lipoprotein metabolism, coagulation, and the production of antiphospholipid autoantibodies. APOH may be a required cofactor for anionic phospholipid binding by the antiphospholipid autoantibodies found in sera of many patients with lupus and primary antiphospholipid syndrome, but it does not seem to be required for the reactivity of antiphospholipid autoantibodies associated with infections.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2GPI; Activated protein C-binding protein; Anticardiolipin cofactor; APC inhibitor; Apo-H; Apolipoprotein H; apolipoprotein H (beta-2-glycoprotein I); B2GPI; beta(2)GPI; Beta-2-glycoprotein 1; Beta-2-glycoprotein I
Gene Aliases: APOH; B2G1; B2GP1; BG
UniProt ID: (Human) P02749
Entrez Gene ID: (Human) 350
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support