Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody detects the 43kD ART5 protein in mouse liver and testis.
Immunogen sequence: MALAALMIA LGSLGLHTWQ AQAVPTILPL GLAPDTFDDT YVGCVEEMEE KAAPLLKEEM AHHALLRESW EAAQETWEDK RRGLTLPPGF KAQNGIAIMV YTNSSNTLYW ELNQAVRTGG GSRELYMRHF PFKALHFYLI RALQLLRGSG GCSRGPGEVV FRGVGSLRFE PKRLGDSVRL GQFASSSLDK AVAHRFGNAT LFSLTTCFGA PIQAFSVFPK EREVLIPPHE VFLVTRFSQD GAQSLVTLWS YNQTCSHFNC AYLGGEKRRG CVSAPGALGT GDLHMTKRHL QQP (1-292 aa encoded by BC014577)
The protein encoded by this gene belongs to the ARG-specific ADP-ribosyltransferase family. Proteins in this family regulate the function of target proteins by attaching ADP-ribose to specific amino acid residues in their target proteins. The mouse homolog lacks a glycosylphosphatidylinositol-anchor signal sequence and is predicted to be a secretory enzyme. Transcript variants with different 5' UTRs, but encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosyltransferase C2 and C3 toxin-like 5; ARTC5; Ecto-ADP-ribosyltransferase 5; Mono(ADP-ribosyl)transferase 5; NAD(P)(+)--arginine ADP-ribosyltransferase 5; YAC-2
Gene Aliases: ART5; ARTC5; UNQ575/PRO1137; Yac-2
UniProt ID: (Human) Q96L15, (Mouse) P70352, (Rat) Q5XHY4
Entrez Gene ID: (Human) 116969, (Mouse) 11875, (Rat) 259167
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support