Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
1 µg/mL of MA5-45453 was sufficient for detection of Cav beta2 in 10 µg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody|Detects approximately 78 kDa. No cross reactivity against Cav beta 1, Cav beta 3, Cav beta 4.
The expression of multiple classes of voltage dependent calcium channels (VDCCs) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCCs exist before birth which is consistent with a role in migration, most N-Type VDCCs subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAB2; CACNLB1; Calcium channel voltage-dependent subunit beta 2; calcium channel, voltage-dependent, beta 2 subunit; CCHLB1; Lambert-Eaton myasthenic syndrome antigen B; MGC138502; MGC138504; MGC41896; myasthenic (Lambert-Eaton) syndrome antigen B; MYSB; Voltage-dependent L-type calcium channel subunit beta-2; voltage-gated calcium channel beta2 subunit transcript variant X13
Gene Aliases: AW060387; CAB2; CACNB2; CACNLB2; CAVB2; Cavbeta2; Cchb2; MYSB
UniProt ID: (Human) Q08289, (Mouse) Q8CC27, (Rat) Q8VGC3
Entrez Gene ID: (Human) 783, (Mouse) 12296, (Rat) 116600
Molecular Function:
voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support