Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SDEEECRIE PCTQKGQCPP PPGLPCPCTG VSDCSGGTDK KLRNCSRLAC LAGELRCTLS DDCIPLTWRC DGHPDCPDSS DELGCGTNEI LPEGDATTMG PPVTLESVTS LRNATTMGPP VTLESVPSVG NATSSSAGDQ SGSPTAYGVI AAAAVLSASL VTATLLLLSW LRAQERLRPL GLLVAMKESL LLSEQKTS (84-280 aa encoded by BC000668 )
Germinal center-B (GC-B) cells differentiate into memory B-cells and plasma cells (PC) through interaction with T-cells and follicular dendritic cells (FDC). CD320 augments the proliferation of PC precursors generated by IL-10. Receptor for the cellular uptake of transcobalamin bound cobalamin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 8D6 antigen; 8D6AFDC-SM-8D6; 8D6TCBLR; CD320; CD320 antigen; FDC-signaling molecule 8D6; FDC-SM-8D6; putative VLDL lipoprotein receptor; TCblR; Transcobalamin receptor
Gene Aliases: 425O18-1; 8D6; 8D6A; CD320; D17Ertd716e; NG29; TCBLR; UNQ198/PRO224; VLDL
UniProt ID: (Human) Q9NPF0, (Mouse) Q9Z1P5
Entrez Gene ID: (Human) 51293, (Mouse) 54219
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support