Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitution: Dissolve the contents of the vial by injection of 0.5 mL sterile distilled water.
Stability Lyophilized product is stable for at least one year at 4°C. After reconstitution, the contents can be safely stored at 4°C for one month or for one year at -20°C.
Add 0.02% sodium azide to prevent bacterial growth
The CD3 subunit complex which is crucial in transducing antigen-recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR complex. T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules and this association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation. The CD3 antigen is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes and Purkinje cells in the cerebellum. It is never expressed on B or NK cells. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases. The genes encoding the CD3 epsilon, gamma and delta polypeptides are located on chromosome 11. Defects in the CD3 gene are associated with CD3 immunodeficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD3 TCR complex; CD3 zeta chain; CD3g molecule, epsilon (CD3-TCR complex); FLJ18683; Leu-4; T cell antigen receptor complex epsilon subunit of T3; T-cell receptor zeta chain; T-cell surface CD3 gamma chain; T3/TCR complex
Gene Aliases: CD3Z; EGK_06950; EGK_06952
Entrez Gene ID: (Rhesus monkey) 697814, (Rhesus monkey) 699582, (Rhesus monkey) 699467, (Rhesus monkey) 705270
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support