Product References
Schlafen14 Impairs HIV-1 Expression in a Codon Usage-Dependent Manner.
Viruses
Valenzuela C,Saucedo S,Llano M
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Mon Mar 25 00:00:00 UTC 2024
Endosialin-positive tumor-derived pericytes promote tumor progression through impeding the infiltration of CD8+ T cells in clear cell renal cell carcinoma.
Cancer immunology, immunotherapy : CII
Lu T,Zhang J,Lu S,Yang F,Gan L,Wu X,Song H,Liu S,Xu C,Han D,Yang B,Wen W,Qin W,Yang L
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Thu Jun 01 00:00:00 UTC 2023
Distinct SARS-CoV-2 specific NLRP3 and IL-1β responses in T cells of aging patients during acute COVID-19 infection.
Frontiers in immunology
Mahalingam SS,Jayaraman S,Arunkumar A,Dudley HM,Anthony DD,Shive CL,Jacobson JM,Pandiyan P
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Thu Oct 12 00:00:00 UTC 2023
Estrogen Protects against Renal Ischemia-Reperfusion Injury by Regulating Th17/Treg Cell Immune Balance.
Disease markers
Zhang Y,Chang Y,Han Z,Ma K,Zeng X,Li L
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Tue Oct 18 00:00:00 UTC 2022
Plasma Extracellular Vesicle Subtypes May be Useful as Potential Biomarkers of Immune Activation in People With HIV.
Pathogens & immunity
Bazié WW,Boucher J,Vitry J,Goyer B,Routy JP,Tremblay C,Trottier S,Jenabian MA,Provost P,Alary M,Gilbert C
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Sat Apr 23 00:00:00 UTC 2022
Targeting Phosphatidylserine Enhances the Anti-tumor Response to Tumor-Directed Radiation Therapy in a Preclinical Model of Melanoma.
Cell reports
Budhu S,Giese R,Gupta A,Fitzgerald K,Zappasodi R,Schad S,Hirschhorn D,Campesato LF,De Henau O,Gigoux M,Liu C,Mazo G,Deng L,Barker CA,Wolchok JD,Merghoub T
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Tue Jan 12 00:00:00 UTC 2021
Immunophenotype of Gastric Tumors Unveils a Pleiotropic Role of Regulatory T Cells in Tumor Development.
Cancers
Rocha S,Basto AP,Ijsselsteijn ME,Teles SP,Azevedo MM,Gonçalves G,Gullo I,Almeida GM,Maqueda JJ,Oliveira MI,Carneiro F,Barata JT,Graça L,de Miranda NFCC,Carvalho J,Oliveira C
11-0048-80 was used in Flow Cytometry to explore the impact of regulatory T cell (Tregs) in gastric cancers (GCs) with distinct histology, and whether Tregs can directly influence tumor cell behavior and GC development.
Sat Jan 23 00:00:00 UTC 2021
Small-molecule antagonist of VLA-4 (GW559090) attenuated neuro-inflammation by targeting Th17 cell trafficking across the blood-retinal barrier in experimental autoimmune uveitis.
Journal of neuroinflammation
Chen YH,Eskandarpour M,Zhang X,Galatowicz G,Greenwood J,Lightman S,Calder V
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Thu Feb 18 00:00:00 UTC 2021
A Cross-Reactive Small Protein Binding Domain Provides a Model to Study Off-Tumor CAR-T Cell Toxicity.
Molecular therapy oncolytics
Hammill JA,Kwiecien JM,Dvorkin-Gheva A,Lau VWC,Baker C,Wu Y,Bezverbnaya K,Aarts C,Heslen CW,Denisova GF,Derocher H,Milne K,Nelson BH,Bramson JL
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Fri Jun 26 00:00:00 UTC 2020
The Potential of Nanobody-Targeted Photodynamic Therapy to Trigger Immune Responses.
Cancers
Beltrán Hernández I,Angelier ML,Del Buono D'Ondes T,Di Maggio A,Yu Y,Oliveira S
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Wed Apr 15 00:00:00 UTC 2020
Expression of CD20 after viral reactivation renders HIV-reservoir cells susceptible to Rituximab.
Nature communications
Serra-Peinado C,Grau-Expósito J,Luque-Ballesteros L,Astorga-Gamaza A,Navarro J,Gallego-Rodriguez J,Martin M,Curran A,Burgos J,Ribera E,Raventós B,Willekens R,Torrella A,Planas B,Badía R,Garcia F,Castellví J,Genescà M,Falcó V,Buzon MJ
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Fri Aug 16 00:00:00 UTC 2019
Accumulation of T-helper 22 cells, interleukin-22 and myeloid-derived suppressor cells promotes gastric cancer progression in elderly patients.
Oncology letters
Chen X,Wang Y,Wang J,Wen J,Jia X,Wang X,Zhang H
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Sun Jul 01 00:00:00 UTC 2018
IL-6 receptor blockade corrects defects of XIAP-deficient regulatory T cells.
Nature communications
Hsieh WC,Hsu TS,Chang YJ,Lai MZ
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Wed Jan 31 00:00:00 UTC 2018
Complement receptor CD46 co-stimulates optimal human CD8+ T cell effector function via fatty acid metabolism.
Nature communications
Arbore G,West EE,Rahman J,Le Friec G,Niyonzima N,Pirooznia M,Tunc I,Pavlidis P,Powell N,Li Y,Liu P,Servais A,Couzi L,Fremeaux-Bacchi V,Placais L,Ferraro A,Walsh PR,Kavanagh D,Afzali B,Lavender P,Lachmann HJ,Kemper C
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Wed Oct 10 00:00:00 UTC 2018
Ex vivo culture of human atherosclerotic plaques: A model to study immune cells in atherogenesis.
Atherosclerosis
Lebedeva A,Vorobyeva D,Vagida M,Ivanova O,Felker E,Fitzgerald W,Danilova N,Gontarenko V,Shpektor A,Vasilieva E,Margolis L
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Fri Dec 01 00:00:00 UTC 2017
Hormonal vitamin D up-regulates tissue-specific PD-L1 and PD-L2 surface glycoprotein expression in humans but not mice.
The Journal of biological chemistry
Dimitrov V,Bouttier M,Boukhaled G,Salehi-Tabar R,Avramescu RG,Memari B,Hasaj B,Lukacs GL,Krawczyk CM,White JH
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Fri Dec 15 00:00:00 UTC 2017
Single-cell profiling reveals GPCR heterogeneity and functional patterning during neuroinflammation.
JCI insight
Tischner D,Grimm M,Kaur H,Staudenraus D,Carvalho J,Looso M,Günther S,Wanke F,Moos S,Siller N,Breuer J,Schwab N,Zipp F,Waisman A,Kurschus FC,Offermanns S,Wettschureck N
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Thu Aug 03 00:00:00 UTC 2017
Targeted Disruption of TCF12 Reveals HEB as Essential in Human Mesodermal Specification and Hematopoiesis.
Stem cell reports
Li Y,Brauer PM,Singh J,Xhiku S,Yoganathan K,Zúñiga-Pflücker JC,Anderson MK
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Tue Sep 12 00:00:00 UTC 2017
CD3xPDL1 bi-specific T cell engager (BiTE) simultaneously activates T cells and NKT cells, kills PDL1+ tumor cells, and extends the survival of tumor-bearing humanized mice.
Oncotarget
Horn LA,Ciavattone NG,Atkinson R,Woldergerima N,Wolf J,Clements VK,Sinha P,Poudel M,Ostrand-Rosenberg S
11-0048 was used in Flow cytometry/Cell sorting to suggest that the CD3xPDL1 BiTE may be efficacious for patients with PDL1(+) solid tumors, in combination with other immunotherapies that do not specifically neutralize PD1 pathway-mediated immune suppression.
Tue Aug 29 00:00:00 UTC 2017
CD3xPDL1 bi-specific T cell engager (BiTE) simultaneously activates T cells and NKT cells, kills PDL1+ tumor cells, and extends the survival of tumor-bearing humanized mice.
Oncotarget
Horn LA,Ciavattone NG,Atkinson R,Woldergerima N,Wolf J,Clements VK,Sinha P,Poudel M,Ostrand-Rosenberg S
11-0048 was used in Flow cytometry/Cell sorting to suggest that the CD3xPDL1 BiTE may be efficacious for patients with PDL1(+) solid tumors, in combination with other immunotherapies that do not specifically neutralize PD1 pathway-mediated immune suppression.
Tue Aug 29 00:00:00 UTC 2017
PD-L1 expression and prognostic impact in glioblastoma.
Neuro-oncology
Nduom EK,Wei J,Yaghi NK,Huang N,Kong LY,Gabrusiewicz K,Ling X,Zhou S,Ivan C,Chen JQ,Burks JK,Fuller GN,Calin GA,Conrad CA,Creasy C,Ritthipichai K,Radvanyi L,Heimberger AB
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Mon Feb 01 00:00:00 UTC 2016
Altered expression of miR-92a correlates with Th17 cell frequency in patients with primary biliary cirrhosis.
International journal of molecular medicine
Liang DY,Hou YQ,Luo LJ,Ao L
11-0048 was used in Flow cytometry/Cell sorting to study the altered expression of miR-92a and its impact on the frequency of Th17 cells in patients with primary biliary cirrhosis.
Fri Jul 01 00:00:00 UTC 2016
Tax and Semaphorin 4D Released from Lymphocytes Infected with Human Lymphotropic Virus Type 1 and Their Effect on Neurite Growth.
AIDS research and human retroviruses
Quintremil S,Alberti C,Rivera M,Medina F,Puente J,Cartier L,Ramírez E,Tanaka Y,Valenzuela MA
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Fri Jan 01 00:00:00 UTC 2016
Tumor-derived exosomes regulate expression of immune function-related genes in human T cell subsets.
Scientific reports
Muller L,Mitsuhashi M,Simms P,Gooding WE,Whiteside TL
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Thu Feb 04 00:00:00 UTC 2016
PSG9 Stimulates Increase in FoxP3+ Regulatory T-Cells through the TGF-β1 Pathway.
PloS one
Jones K,Ballesteros A,Mentink-Kane M,Warren J,Rattila S,Malech H,Kang E,Dveksler G
11-0048 was used in Flow cytometry/Cell sorting to investigate whether pregnancy-specific glycoprotein 9, a potential predictive biomarker of pre-eclampsia, plays an immunoregulatory role during pregnancy.
Wed Jul 26 00:00:00 UTC 2017
The Expression of Programmed Death-1 on CD4+ and CD8+ T Lymphocytes in Patients with Type 2 Diabetes and Severe Sepsis.
PloS one
Jia Y,Zhao Y,Li C,Shao R
11-0048 was used in Flow cytometry/Cell sorting to investigate the expression of Programmed death-1 on T lymphocytes in patients with type 2 diabetes mellitus and severe sepsis.
Mon Jul 24 00:00:00 UTC 2017
A Systematic Study of the Effect of Different Molecular Weights of Hyaluronic Acid on Mesenchymal Stromal Cell-Mediated Immunomodulation.
PloS one
Gómez-Aristizábal A,Kim KP,Viswanathan S
11-0048 was used in Flow cytometry/Cell sorting to investigate the combined mechanism of action of MSCs and hyaluronic acid.
Wed Jul 06 00:00:00 UTC 2016
Homeostatic regulation of T cell trafficking by a B cell-derived peptide is impaired in autoimmune and chronic inflammatory disease.
Nature medicine
Chimen M,McGettrick HM,Apta B,Kuravi SJ,Yates CM,Kennedy A,Odedra A,Alassiri M,Harrison M,Martin A,Barone F,Nayar S,Hitchcock JR,Cunningham AF,Raza K,Filer A,Copland DA,Dick AD,Robinson J,Kalia N,Walker LSK,Buckley CD,Nash GB,Narendran P,Rainger GE
11-0048 was used in Flow cytometry/Cell sorting to demonstrate that exogenous PEPITEM reduces T cell recruitment into inflamed tissues in multiple animal models of autoimmune, inflammatory diseases.
Fri May 01 00:00:00 UTC 2015
Low B and T lymphocyte attenuator expression on CD4+ T cells in the early stage of sepsis is associated with the severity and mortality of septic patients: a prospective cohort study.
Critical care (London, England)
Shao R,Li CS,Fang Y,Zhao L,Hang C
11-0048 was used in Flow cytometry/Cell sorting to investigate B and T lymphocyte attenuator expression on CD4(+) T cells in healthy controls and septic patients.
Fri Aug 28 00:00:00 UTC 2015
The HIV-1 gp120 CD4-bound conformation is preferentially targeted by antibody-dependent cellular cytotoxicity-mediating antibodies in sera from HIV-1-infected individuals.
Journal of virology
Veillette M,Coutu M,Richard J,Batraville LA,Dagher O,Bernard N,Tremblay C,Kaufmann DE,Roger M,Finzi A
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Thu Jan 01 00:00:00 UTC 2015
Skewed T-helper (Th)1/2- and Th17/T regulatory‑cell balances in patients with renal cell carcinoma.
Molecular medicine reports
Li L,Yang C,Zhao Z,Xu B,Zheng M,Zhang C,Min Z,Guo J,Rong R
11-0048 was used in Flow cytometry/Cell sorting to demonstrate that the balance of Th1 and Th2 cells is skewed towards the Th2 profile in the peripheral blood of patients with RCC.
Sun Feb 01 00:00:00 UTC 2015
Aging and cytomegalovirus infection differentially and jointly affect distinct circulating T cell subsets in humans.
Journal of immunology (Baltimore, Md. : 1950)
Wertheimer AM,Bennett MS,Park B,Uhrlaub JL,Martinez C,Pulko V,Currier NL,Nikolich-Žugich D,Kaye J,Nikolich-Žugich J
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Flow Cytometry
Sat Mar 01 00:00:00 UTC 2014
Calcium signaling via Orai1 is essential for induction of the nuclear orphan receptor pathway to drive Th17 differentiation.
Journal of immunology (Baltimore, Md. : 1950)
Kim KD,Srikanth S,Tan YV,Yee MK,Jew M,Damoiseaux R,Jung ME,Shimizu S,An DS,Ribalet B,Waschek JA,Gwack Y
11-0048 was used in Flow cytometry/Cell sorting to identify a novel class of small molecules that inhibit Ca(2+) release-activated Ca(2+) channel activity.
Wed Jan 01 00:00:00 UTC 2014
An increased abundance of tumor-infiltrating regulatory T cells is correlated with the progression and prognosis of pancreatic ductal adenocarcinoma.
PloS one
Tang Y,Xu X,Guo S,Zhang C,Tang Y,Tian Y,Ni B,Lu B,Wang H
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Immunohistochemistry
Tue Jan 06 00:00:00 UTC 2015
An increased abundance of tumor-infiltrating regulatory T cells is correlated with the progression and prognosis of pancreatic ductal adenocarcinoma.
PloS one
Tang Y,Xu X,Guo S,Zhang C,Tang Y,Tian Y,Ni B,Lu B,Wang H
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Immunohistochemistry
Tue Jan 06 00:00:00 UTC 2015
Azithromycin suppresses CD4(+) T-cell activation by direct modulation of mTOR activity.
Scientific reports
Ratzinger F,Haslacher H,Poeppl W,Hoermann G,Kovarik JJ,Jutz S,Steinberger P,Burgmann H,Pickl WF,Schmetterer KG
11-0048 was used in Flow cytometry/Cell sorting to evaluate the in vitro effects of the antibiotics azithromycin and clarithromycin on the activation of CD4(+) T cells.
Thu Dec 11 00:00:00 UTC 2014
Association of a NOD2 gene polymorphism and T-helper 17 cells with presumed ocular toxoplasmosis.
The Journal of infectious diseases
Dutra MS,Béla SR,Peixoto-Rangel AL,Fakiola M,Cruz AG,Gazzinelli A,Quites HF,Bahia-Oliveira LM,Peixe RG,Campos WR,Higino-Rocha AC,Miller NE,Blackwell JM,Antonelli LR,Gazzinelli RT
11-0048 was used in Flow cytometry/Cell sorting to suggest that NOD2 influences the production of IL-17A by CD4 T lymphocytes and might contribute to the development of ocular toxoplasmosis.
Tue Jan 01 00:00:00 UTC 2013
Memory T cells in latent Mycobacterium tuberculosis infection are directed against three antigenic islands and largely contained in a CXCR3+CCR6+ Th1 subset.
PLoS pathogens
Lindestam Arlehamn CS,Gerasimova A,Mele F,Henderson R,Swann J,Greenbaum JA,Kim Y,Sidney J,James EA,Taplitz R,McKinney DM,Kwok WW,Grey H,Sallusto F,Peters B,Sette A
11-0048 was used in Flow cytometry/Cell sorting to further define the immunological footprint of Mycobacterium tuberculosis CD4 T cell recognition.
Tue Jan 01 00:00:00 UTC 2013
Role of gastric epithelial cell-derived transforming growth factor beta in reduced CD4+ T cell proliferation and development of regulatory T cells during Helicobacter pylori infection.
Infection and immunity
Beswick EJ,Pinchuk IV,Earley RB,Schmitt DA,Reyes VE
11-0048 was used in Flow cytometry/Cell sorting to show how TGF-β produced by gastric epithelial cells inhibits CD4+ T cell responses during H. pylori infection, suggesting a possible mechanism of persistence.
Fri Jul 01 00:00:00 UTC 2011
Common gamma chain cytokines promote rapid in vitro expansion of allo-specific human CD8+ suppressor T cells.
PloS one
Yu Y,Zitzner JR,Houlihan J,Herrera N,Xu L,Miller J,Mathew JM,Tambur AR,Luo X
11-0048 was used in Flow cytometry/Cell sorting to demonstrate the use of common γc cytokines in combination with donor APCs as a strategy for generation of donor-specific T cells for immunotherapy.
Mon Aug 27 00:00:00 UTC 2012
Assessment of seasonal influenza A virus-specific CD4 T-cell responses to 2009 pandemic H1N1 swine-origin influenza A virus.
Journal of virology
Ge X,Tan V,Bollyky PL,Standifer NE,James EA,Kwok WW
11-0048 was used in Flow cytometry/Cell sorting to investigate how, without protective antibody responses, individuals vaccinated against seasonal influenza A may still benefit from preexisting cross-reactive memory CD4 T cells reducing their susceptibility to S-OIV infection.
Thu Apr 01 00:00:00 UTC 2010
CD4-CCR5 interaction in intracellular compartments contributes to receptor expression at the cell surface.
Blood
Achour L,Scott MG,Shirvani H,Thuret A,Bismuth G,Labbé-Jullié C,Marullo S
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Western Blot
Thu Feb 26 00:00:00 UTC 2009
CD4-CCR5 interaction in intracellular compartments contributes to receptor expression at the cell surface.
Blood
Achour L,Scott MG,Shirvani H,Thuret A,Bismuth G,Labbé-Jullié C,Marullo S
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Western Blot
Thu Feb 26 00:00:00 UTC 2009
CD4-CCR5 interaction in intracellular compartments contributes to receptor expression at the cell surface.
Blood
Achour L,Scott MG,Shirvani H,Thuret A,Bismuth G,Labbé-Jullié C,Marullo S
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Western Blot
Thu Feb 26 00:00:00 UTC 2009
CD4-CCR5 interaction in intracellular compartments contributes to receptor expression at the cell surface.
Blood
Achour L,Scott MG,Shirvani H,Thuret A,Bismuth G,Labbé-Jullié C,Marullo S
Published figure using CD4 monoclonal antibody (Product # 11-0048-80) in Western Blot
Thu Feb 26 00:00:00 UTC 2009
Naive CD4 t cell proliferation is controlled by mammalian target of rapamycin regulation of GRAIL expression.
Journal of immunology (Baltimore, Md. : 1950)
Lin JT,Lineberry NB,Kattah MG,Su LL,Utz PJ,Fathman CG,Wu L
11-0048 was used in Flow cytometry/Cell sorting to investigate the role of GRAIL in CD4 T cells, showing that proliferation is controlled by mTOR regulation of GRAIL expression.
Fri May 15 00:00:00 UTC 2009
T-cell responses to survivin in cancer patients undergoing radiation therapy.
Clinical cancer research : an official journal of the American Association for Cancer Research
Schaue D,Comin-Anduix B,Ribas A,Zhang L,Goodglick L,Sayre JW,Debucquoy A,Haustermans K,McBride WH
11-0048 was used in Flow cytometry/Cell sorting to show that radiation therapy may increase the likelihood of some cancer patients responding to immunotherapy.
Fri Aug 01 00:00:00 UTC 2008
TCR-induced downregulation of protein tyrosine phosphatase PEST augments secondary T cell responses.
Molecular immunology
Arimura Y,Vang T,Tautz L,Williams S,Mustelin T
11-0048 was used in Flow cytometry/Cell sorting to investigate how PTP-PEST is down-regulated following T cell activation, and how that can lead to reduced IL-2 secretion following TCR and CD28 stimulation.
Sun Jun 01 00:00:00 UTC 2008
T regulatory cell levels decrease in people infected with Schistosoma mansoni on effective treatment.
The American journal of tropical medicine and hygiene
Watanabe K,Mwinzi PN,Black CL,Muok EM,Karanja DM,Secor WE,Colley DG
11-0048 was used in Flow cytometry/Cell sorting to investigate the role of Tregs in Schistosoma mansoni infection, showing that Treg levels decrease in people on effective treatment.
Mon Oct 01 00:00:00 UTC 2007