Product References
A single-cell transcriptional atlas reveals resident progenitor cell niche functions in TMJ disc development and injury.
Nature communications
Bi R,Yin Q,Li H,Yang X,Wang Y,Li Q,Fang H,Li P,Lyu P,Fan Y,Ying B,Zhu S
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 17-0902-82) in Flow Cytometry
Tue Feb 14 00:00:00 EST 2023
Airway Administration of Bacterial Lysate OM-85 Protects Mice Against Respiratory Syncytial Virus Infection.
Frontiers in immunology
Antunes KH,Cassão G,Santos LD,Borges SG,Poppe J,Gonçalves JB,Nunes EDS,Recacho GF,Sousa VB,Da Silva GS,Mansur D,Stein RT,Pasquali C,De Souza APD
17-0902-82 was used in Flow cytometry/Cell sorting to suggest that intranasal use of OM-85 should be considered as a potential prophylactic product to prevent RSV bronchiolitis once human studies confirm these findings.
Tue May 24 00:00:00 EDT 2022
Pannexin 1 channels facilitate communication between T cells to restrict the severity of airway inflammation.
Immunity
Medina CB,Chiu YH,Stremska ME,Lucas CD,Poon I,Tung KS,Elliott MR,Desai B,Lorenz UM,Bayliss DA,Ravichandran KS
17-0902-82 was used in Flow cytometry/Cell sorting to uncover an immunoregulatory role for the nucleotide release channel, Panx1, in T cell crosstalk during airway disease.
Tue Aug 10 00:00:00 EDT 2021
Therapeutic Potential of Mesenchymal Stem Cells in a Pre-Clinical Model of Diabetic Kidney Disease and Obesity.
International journal of molecular sciences
Sávio-Silva C,Soinski-Sousa PE,Simplício-Filho A,Bastos RMC,Beyerstedt S,Rangel ÉB
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 17-0902-82) in Flow Cytometry
Thu Feb 04 00:00:00 EST 2021
Quorum Regulation via Nested Antagonistic Feedback Circuits Mediated by the Receptors CD28 and CTLA-4 Confers Robustness to T Cell Population Dynamics.
Immunity
Zenke S,Palm MM,Braun J,Gavrilov A,Meiser P,Böttcher JP,Beyersdorf N,Ehl S,Gerard A,Lämmermann T,Schumacher TN,Beltman JB,Rohr JC
17-0902 was used in Flow cytometry/Cell sorting to conclude that CD8+ T cell-population-intrinsic mechanisms regulate cellular behavior, thereby promoting robustness of population dynamics.
Tue Feb 18 00:00:00 EST 2020
Extracellular HMGB-1 activates inflammatory signaling in tendon cells and tissues.
Therapeutic advances in chronic disease
Zhang C,Gu X,Zhao G,Wang W,Shao J,Zhu J,Yuan T,Sun J,Nie D,Zhou Y
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 17-0902-82) in Flow Cytometry
Sun Apr 17 00:00:00 EDT 2022
Loss of Fas signaling in fibroblasts impairs homeostatic fibrosis resolution and promotes persistent pulmonary fibrosis.
JCI insight
Redente EF,Chakraborty S,Sajuthi S,Black BP,Edelman BL,Seibold MA,Riches DW
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 17-0902-82) in Flow Cytometry
Tue Dec 08 00:00:00 EST 2020
Neurokinin-1 Receptor Signaling Is Required for Efficient Ca2+ Flux in T-Cell-Receptor-Activated T Cells.
Cell reports
Morelli AE,Sumpter TL,Rojas-Canales DM,Bandyopadhyay M,Chen Z,Tkacheva O,Shufesky WJ,Wallace CT,Watkins SC,Berger A,Paige CJ,Falo LD,Larregina AT
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 17-0902-82) in Flow Cytometry
Tue Mar 10 00:00:00 EDT 2020
Cardiac fibroblast proliferation rates and collagen expression mature early and are unaltered with advancing age.
JCI insight
Wu R,Ma F,Tosevska A,Farrell C,Pellegrini M,Deb A
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 17-0902-82) in Flow Cytometry
Thu Dec 17 00:00:00 EST 2020
Cardiac fibroblast proliferation rates and collagen expression mature early and are unaltered with advancing age.
JCI insight
Wu R,Ma F,Tosevska A,Farrell C,Pellegrini M,Deb A
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 17-0902-82) in Flow Cytometry
Thu Dec 17 00:00:00 EST 2020
Critical role of WNK1 in MYC-dependent early mouse thymocyte development.
eLife
Köchl R,Vanes L,Llorian Sopena M,Chakravarty P,Hartweger H,Fountain K,White A,Cowan J,Anderson G,Tybulewicz VL
17-0902 was used in Flow cytometry/Cell sorting to ind that WNK1 is required for the transition of double negative (DN) thymocytes through the β-selection checkpoint and subsequent proliferation and differentiation into double positive (DP) thymocytes.
Wed Oct 14 00:00:00 EDT 2020
Discovery of a periosteal stem cell mediating intramembranous bone formation.
Nature
Debnath S,Yallowitz AR,McCormick J,Lalani S,Zhang T,Xu R,Li N,Liu Y,Yang YS,Eiseman M,Shim JH,Hameed M,Healey JH,Bostrom MP,Landau DA,Greenblatt MB
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 17-0902-82) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2018
The Severe Deficiency of the Somatotrope GH-Releasing Hormone/Growth Hormone/Insulin-Like Growth Factor 1 Axis of Mice Is Associated With an Important Splenic Atrophy and Relative B Lymphopenia.
Frontiers in endocrinology
Bodart G,Farhat K,Renard-Charlet C,Becker G,Plenevaux A,Salvatori R,Geenen V,Martens H
17-0902 was used in Flow cytometry/Cell sorting to examine the precise control exerted by the somatotrope growth hormone(GH)-releasing hormone/GH/insulin-like growth factor 1 axis on the immune system.
Sun Apr 10 00:00:00 EDT 2022
Precardiac organoids form two heart fields via Bmp/Wnt signaling.
Nature communications
Andersen P,Tampakakis E,Jimenez DV,Kannan S,Miyamoto M,Shin HK,Saberi A,Murphy S,Sulistio E,Chelko SP,Kwon C
17-0902 was used in Immunohistochemistry to study the specification of two cardiac origins via Bmp/Wnt signalling.
Tue Aug 07 00:00:00 EDT 2018
Group 2 innate lymphoid cells protect lung endothelial cells from pyroptosis in sepsis.
Cell death & disease
Lai D,Tang J,Chen L,Fan EK,Scott MJ,Li Y,Billiar TR,Wilson MA,Fang X,Shu Q,Fan J
17-0902 was used in Flow cytometry/Cell sorting to describe the function of ILC2 in the regulation of lung inflammation by protecting lung endothelial cell from pyroptosis.
Tue Mar 06 00:00:00 EST 2018
Single-cell transcriptomics reconstructs fate conversion from fibroblast to cardiomyocyte.
Nature
Liu Z,Wang L,Welch JD,Ma H,Zhou Y,Vaseghi HR,Yu S,Wall JB,Alimohamadi S,Zheng M,Yin C,Shen W,Prins JF,Liu J,Qian L
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 17-0902-82) in Flow Cytometry
Thu Nov 02 00:00:00 EDT 2017
Mutations in 5-methylcytosine oxidase TET2 and RhoA cooperatively disrupt T cell homeostasis.
The Journal of clinical investigation
Zang S,Li J,Yang H,Zeng H,Han W,Zhang J,Lee M,Moczygemba M,Isgandarova S,Yang Y,Zhou Y,Rao A,You MJ,Sun D,Huang Y
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 17-0902-82) in Flow Cytometry
Tue Aug 01 00:00:00 EDT 2017
CTCF-Mediated Enhancer-Promoter Interaction Is a Critical Regulator of Cell-to-Cell Variation of Gene Expression.
Molecular cell
Ren G,Jin W,Cui K,Rodrigez J,Hu G,Zhang Z,Larson DR,Zhao K
Thu Sep 21 00:00:00 EDT 2017
A population of hematopoietic stem cells derives from GATA4-expressing progenitors located in the placenta and lateral mesoderm of mice.
Haematologica
Cañete A,Carmona R,Ariza L,Sánchez MJ,Rojas A,Muñoz-Chápuli R
17090282 was used in flow cytometry to utilize a G2-Gata4Cre;R26REYFP mouse line to track the developmental fate of the G2-Gata4 cell lineage
Sat Apr 01 00:00:00 EDT 2017
Endocardium Minimally Contributes to Coronary Endothelium in the Embryonic Ventricular Free Walls.
Circulation research
Zhang H,Pu W,Li G,Huang X,He L,Tian X,Liu Q,Zhang L,Wu SM,Sucov HM,Zhou B
17-0902 was used in Flow cytometry/Cell sorting to determine through genetic lineage tracing that sinus venosus is the major origin for coronary endothelium in the embryonic ventricular free walls.
Fri Jun 10 00:00:00 EDT 2016
Lymphatic Contribution to the Cellular Niche in Heterotopic Ossification.
Annals of surgery
Loder S,Agarwal S,Sorkin M,Breuler C,Li J,Peterson J,Gardenier J,Hsieh HH,Wang SC,Mehrara BJ,Levi B
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 17-0902-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2016
Survival of Igα-Deficient Mature B Cells Requires BAFF-R Function.
Journal of immunology (Baltimore, Md. : 1950)
Levit-Zerdoun E,Becker M,Pohlmeyer R,Wilhelm I,Maity PC,Rajewsky K,Reth M,Hobeika E
17-0902 was used in Flow cytometry/Cell sorting to assess the importance of the expression of a functional B-cell receptor for B cell maintenance.
Tue Mar 01 00:00:00 EST 2016
Heme Oxygenase-1-Expressing Dendritic Cells Promote Foxp3+ Regulatory T Cell Differentiation and Induce Less Severe Airway Inflammation in Murine Models.
PloS one
Wong TH,Chen HA,Gau RJ,Yen JH,Suen JL
17-0902 was used in Flow cytometry/Cell sorting to study the role of heme oxygenase-1 in dendritic cell-mediated T cell differentiation.
Mon Jul 10 00:00:00 EDT 2017
Injectable, spontaneously assembling, inorganic scaffolds modulate immune cells in vivo and increase vaccine efficacy.
Nature biotechnology
Kim J,Li WA,Choi Y,Lewin SA,Verbeke CS,Dranoff G,Mooney DJ
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 17-0902-82) in Flow Cytometry
Thu Jan 01 00:00:00 EST 2015
MicroRNA-451 exacerbates lipotoxicity in cardiac myocytes and high-fat diet-induced cardiac hypertrophy in mice through suppression of the LKB1/AMPK pathway.
Circulation research
Kuwabara Y,Horie T,Baba O,Watanabe S,Nishiga M,Usami S,Izuhara M,Nakao T,Nishino T,Otsu K,Kita T,Kimura T,Ono K
17-0902 was used in Flow cytometry/Cell sorting to elucidate the mechanisms underlying the involvement of miR-451 in diabetic cardiomyopathy.
Fri Jan 16 00:00:00 EST 2015
Mouse bone marrow-derived mesenchymal stem cells inhibit leukemia/lymphoma cell proliferation in vitro and in a mouse model of allogeneic bone marrow transplant.
International journal of molecular medicine
Song N,Gao L,Qiu H,Huang C,Cheng H,Zhou H,Lv S,Chen L,Wang J
17-0902 was used in Flow cytometry/Cell sorting to determine that MSCs may be safe and effective when used in allogeneic BMT for the treatment of haemotological malignancies.
Wed Jul 01 00:00:00 EDT 2015
Disruption of p21-activated kinase 1 gene diminishes atherosclerosis in apolipoprotein E-deficient mice.
Nature communications
Singh NK,Kotla S,Dyukova E,Traylor JG,Orr AW,Chernoff J,Marion TN,Rao GN
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 17-0902-82) in Flow Cytometry
Wed Jun 24 00:00:00 EDT 2015
Expression Patterns of miRNA-423-5p in the Serum and Pericardial Fluid in Patients Undergoing Cardiac Surgery.
PloS one
Miyamoto S,Usami S,Kuwabara Y,Horie T,Baba O,Hakuno D,Nakashima Y,Nishiga M,Izuhara M,Nakao T,Nishino T,Ide Y,Nakazeki F,Wang J,Ueyama K,Kimura T,Ono K
17-0902 was used in Flow cytometry/Cell sorting to suggested that miR-423-5p is enriched in PF, and serum miR-423-5p may be associated with uAP.
Wed Jul 06 00:00:00 EDT 2016
Interleukin-17-producing innate lymphoid cells and the NLRP3 inflammasome facilitate obesity-associated airway hyperreactivity.
Nature medicine
Kim HY,Lee HJ,Chang YJ,Pichavant M,Shore SA,Fitzgerald KA,Iwakura Y,Israel E,Bolger K,Faul J,DeKruyff RH,Umetsu DT
17-0902 was used in Flow cytometry/Cell sorting to show that IL-17 producing innate lymphoid cells and the NLRP3 inflammasome facilitate obesity-associated airway hyperreactivity.
Wed Jan 01 00:00:00 EST 2014
C/EBPδ deficiency sensitizes mice to ionizing radiation-induced hematopoietic and intestinal injury.
PloS one
Pawar SA,Shao L,Chang J,Wang W,Pathak R,Zhu X,Wang J,Hendrickson H,Boerma M,Sterneck E,Zhou D,Hauer-Jensen M
17-0902 was used in Flow cytometry/Cell sorting to uncover a novel biological function for C/EBPδ in promoting the response to radiation-induced DNA-damage and in protecting hematopoietic and intestinal tissues from radiation-induced injury.
Thu Jan 15 00:00:00 EST 2015
Mutations in Alström protein impair terminal differentiation of cardiomyocytes.
Nature communications
Shenje LT,Andersen P,Halushka MK,Lui C,Fernandez L,Collin GB,Amat-Alarcon N,Meschino W,Cutz E,Chang K,Yonescu R,Batista DA,Chen Y,Chelko S,Crosson JE,Scheel J,Vricella L,Craig BD,Marosy BA,Mohr DW,Hetrick KN,Romm JM,Scott AF,Valle D,Naggert JK,Kwon C,Doheny KF,Judge DP
17-0902 was used in Flow cytometry/Cell sorting to conclude that deficiency of Alström protein impairs postnatal cardiomyocyte cell cycle arrest.
Tue Mar 04 00:00:00 EST 2014
High-resolution analysis with novel cell-surface markers identifies routes to iPS cells.
Nature
O'Malley J,Skylaki S,Iwabuchi KA,Chantzoura E,Ruetz T,Johnsson A,Tomlinson SR,Linnarsson S,Kaji K
17-0902 was used in Flow cytometry/Cell sorting to investigate the routes that fibroblasts take during the generation of induced pluripotent stem cells, showing that there are orderly changes in cell-surface markers.
Thu Jul 04 00:00:00 EDT 2013
Themis sets the signal threshold for positive and negative selection in T-cell development.
Nature
Fu G,Casas J,Rigaud S,Rybakin V,Lambolez F,Brzostek J,Hoerter JA,Paster W,Acuto O,Cheroutre H,Sauer K,Gascoigne NR
17-0902 was used in Flow cytometry/Cell sorting to show that the T-cell-specific protein Themis can specifically attenuate TCR signal strength via SHP1 recruitment and activation in response to low-affinity TCR engagement.
Thu Dec 19 00:00:00 EST 2013
Interleukin-33-dependent innate lymphoid cells mediate hepatic fibrosis.
Immunity
McHedlidze T,Waldner M,Zopf S,Walker J,Rankin AL,Schuchmann M,Voehringer D,McKenzie AN,Neurath MF,Pflanz S,Wirtz S
17-0902 was used in Flow cytometry/Cell sorting to investigate the role of inflammatory networks in liver fibrosis, showing that it is mediated by interleukin-33-dependent innate lymphoid cells.
Thu Aug 22 00:00:00 EDT 2013
Notch2-dependent classical dendritic cells orchestrate intestinal immunity to attaching-and-effacing bacterial pathogens.
Nature immunology
Satpathy AT,Briseño CG,Lee JS,Ng D,Manieri NA,Kc W,Wu X,Thomas SR,Lee WL,Turkoz M,McDonald KG,Meredith MM,Song C,Guidos CJ,Newberry RD,Ouyang W,Murphy TL,Stappenbeck TS,Gommerman JL,Nussenzweig MC,Colonna M,Kopan R,Murphy KM
17-0902 was used in Flow cytometry/Cell sorting to elucidate the cellular source of essential IL-23 production required for the mucosal response to attaching-and-effacing bacterial infection.
Sun Sep 01 00:00:00 EDT 2013
Absence of signaling into CD4⁺ cells via C3aR and C5aR enables autoinductive TGF-β1 signaling and induction of Foxp3⁺ regulatory T cells.
Nature immunology
Strainic MG,Shevach EM,An F,Lin F,Medof ME
17-0902 was used in Flow cytometry/Cell sorting to demonstrate that a lack of signalling can convert CD4(+) cells to Foxp3(+) iT(reg) cells, amongst other effects.
Fri Feb 01 00:00:00 EST 2013
Regulatory T cells that recognize a ubiquitous stress-inducible self-antigen are long-lived suppressors of autoimmune arthritis.
Proceedings of the National Academy of Sciences of the United States of America
van Herwijnen MJ,Wieten L,van der Zee R,van Kooten PJ,Wagenaar-Hilbers JP,Hoek A,den Braber I,Anderton SM,Singh M,Meiring HD,van Els CA,van Eden W,Broere F
17-0902 was used in Flow cytometry/Cell sorting to uncover potent disease-suppressive Treg cells that recognise Hsp 70, with the potential for re-establishing self-tolerance in autoimmunity.
Tue Aug 28 00:00:00 EDT 2012
Biological and clinical availability of adipose-derived stem cells for pelvic dead space repair.
Stem cells translational medicine
Takahashi H,Haraguchi N,Nishikawa S,Miyazaki S,Suzuki Y,Mizushima T,Nishimura J,Takemasa I,Yamamoto H,Mimori K,Ishii H,Doki Y,Mori M
17-0902 was used in Flow cytometry/Cell sorting to clarify the utility of adipose-derived stem cells and resected intraperitoneal fat pads for pelvic cavity repair using a mouse model.
Thu Nov 01 00:00:00 EDT 2012
T cell apoptosis and induction of Foxp3+ regulatory T cells underlie the therapeutic efficacy of CD4 blockade in experimental autoimmune encephalomyelitis.
Journal of immunology (Baltimore, Md. : 1950)
Duarte J,Carrié N,Oliveira VG,Almeida C,Agua-Doce A,Rodrigues L,Simas JP,Mars LT,Graca L
17-0902 was used in Flow cytometry/Cell sorting to investigate the mechanism underlying effective MS prevention, showing that apoptosis of T cells and induction of Foxp3+ Tregs underlie efficacy of CD4 blockade in EAE.
Wed Aug 15 00:00:00 EDT 2012
Partial characterization of the Sox2+ cell population in an adult murine model of digit amputation.
Tissue engineering. Part A
Agrawal V,Siu BF,Chao H,Hirschi KK,Raborn E,Johnson SA,Tottey S,Hurley KB,Medberry CJ,Badylak SF
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 17-0902-82) in Flow Cytometry
Sun Jul 01 00:00:00 EDT 2012
Deleterious effect of the IL-23/IL-17A axis and γδT cells on left ventricular remodeling after myocardial infarction.
Journal of the American Heart Association
Yan X,Shichita T,Katsumata Y,Matsuhashi T,Ito H,Ito K,Anzai A,Endo J,Tamura Y,Kimura K,Fujita J,Shinmura K,Shen W,Yoshimura A,Fukuda K,Sano M
17-0902 was used in Flow cytometry/Cell sorting to explore potential immune pathways involved in left ventricular remodelling as possible preventative therapeutic options.
Mon Oct 01 00:00:00 EDT 2012
Nuclear export of the NF-κB inhibitor IκBα is required for proper B cell and secondary lymphoid tissue formation.
Immunity
Wuerzberger-Davis SM,Chen Y,Yang DT,Kearns JD,Bates PW,Lynch C,Ladell NC,Yu M,Podd A,Zeng H,Huang TT,Wen R,Hoffmann A,Wang D,Miyamoto S
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 17-0902-82) in Flow Cytometry
Fri Feb 25 00:00:00 EST 2011
Vitamin A deficiency impairs vaccine-elicited gastrointestinal immunity.
Journal of immunology (Baltimore, Md. : 1950)
Kaufman DR,De Calisto J,Simmons NL,Cruz AN,Villablanca EJ,Mora JR,Barouch DH
17-0902 was used in Flow cytometry/Cell sorting to suggest that oral vitamin A supplementation may be important for optimizing the success of vaccines against HIV-1 and other mucosal pathogens in the developing world.
Mon Aug 15 00:00:00 EDT 2011
A critical role of TAK1 in B-cell receptor-mediated nuclear factor kappaB activation.
Blood
Schuman J,Chen Y,Podd A,Yu M,Liu HH,Wen R,Chen ZJ,Wang D
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 17-0902-82) in Flow Cytometry
Thu May 07 00:00:00 EDT 2009
Tracking epitope-specific T cells.
Nature protocols
Moon JJ,Chu HH,Hataye J,Pagán AJ,Pepper M,McLachlan JB,Zell T,Jenkins MK
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 17-0902-82) in Flow Cytometry
Tue Jun 30 00:00:00 EDT 2009
Trans-presentation of IL-15 by intestinal epithelial cells drives development of CD8alphaalpha IELs.
Journal of immunology (Baltimore, Md. : 1950)
Ma LJ,Acero LF,Zal T,Schluns KS
17-0902 was used in Flow cytometry/Cell sorting to identify the cell type responsible for the delivery of IL-15 to intraepithelial lymphocytes.
Wed Jul 15 00:00:00 EDT 2009
Autoantibody production in lpr/lpr gld/gld mice reflects accumulation of CD4+ effector cells that are resistant to regulatory T cell activity.
Journal of autoimmunity
Hondowicz BD,Fields ML,Nish SA,Larkin J,Caton AJ,Erikson J
17-0902 was used in Flow cytometry/Cell sorting to show that Treg cells block lpr/lpr gld/gld Th cells from providing help to anti-chromatin B cells in an in vivo transfer system.
Mon Sep 01 00:00:00 EDT 2008
Manganese superoxide dismutase gene dosage affects chromosomal instability and tumor onset in a mouse model of T cell lymphoma.
Free radical biology & medicine
van de Wetering CI,Coleman MC,Spitz DR,Smith BJ,Knudson CM
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 17-0902-82) in Flow Cytometry
Tue Apr 15 00:00:00 EDT 2008
The crystal structure of CD8 in complex with YTS156.7.7 Fab and interaction with other CD8 antibodies define the binding mode of CD8 alphabeta to MHC class I.
Journal of molecular biology
Shore DA,Issafras H,Landais E,Teyton L,Wilson IA
17-0902 was used in Flow cytometry/Cell sorting to characterise the interaction between CD8alphabeta heterodimer and class I pMHC on antigen-presenting cells.
Wed Dec 31 00:00:00 EST 2008
Dynamics of lung macrophage activation in response to helminth infection.
Journal of leukocyte biology
Siracusa MC,Reece JJ,Urban JF,Scott AL
17-0902 was used in Flow cytometry/Cell sorting to characterise the surface molecules, gene expression, and cell function of macrophages isolated from lung tissue after helminth infection.
Mon Dec 01 00:00:00 EST 2008
Activated CD8 T cells redistribute to antigen-free lymph nodes and exhibit effector and memory characteristics.
Journal of immunology (Baltimore, Md. : 1950)
Brinkman CC,Sheasley-O'Neill SL,Ferguson AR,Engelhard VH
17-0902 was used in Flow cytometry/Cell sorting to characterise the activation and differentiation of CD8 T cells in individual lymphoid compartments over time, and their migration between compartments.
Fri Aug 01 00:00:00 EDT 2008
Homeostasis and effector function of lymphopenia-induced "memory-like" T cells in constitutively T cell-depleted mice.
Journal of immunology (Baltimore, Md. : 1950)
Voehringer D,Liang HE,Locksley RM
17-0902-82 was used in Flow Cytometry to demonstrate that T(ML) cells are not sufficient to mediate an effective immune response in constitutively T cell-depleted mice.
Tue Apr 01 00:00:00 EDT 2008
Tumor growth impedes natural-killer-cell maturation in the bone marrow.
Blood
Richards JO,Chang X,Blaser BW,Caligiuri MA,Zheng P,Liu Y
17-0902 was used in Flow cytometry/Cell sorting to demonstrate that tumour growth can impede functional maturation of NK cells, most likely by interrupting the requisite IL-15 signalling pathway.
Sat Jul 01 00:00:00 EDT 2006
Converting IL-15 to a superagonist by binding to soluble IL-15R{alpha}.
Proceedings of the National Academy of Sciences of the United States of America
Rubinstein MP,Kovar M,Purton JF,Cho JH,Boyman O,Surh CD,Sprent J
17-0902 was used in Flow cytometry/Cell sorting to study how IL-2 function is markedly inhibited by binding to soluble IL-2Ralpha.
Tue Jun 13 00:00:00 EDT 2006
An MHC-linked locus modulates thymic differentiation of CD4+CD25+Foxp3+ regulatory T lymphocytes.
International immunology
Tellier J,van Meerwijk JP,Romagnoli P
17-0902 was used in Flow cytometry/Cell sorting to investigate factors that affect the levels of Tregs in the thymus.
Wed Nov 01 00:00:00 EST 2006