Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: PEKPVNISCW SKNMKDLTCR WTPGAHGETF LHTNYSLKYK LRWYGQDNTC EEYHTVGPHS CHIPKDLALF TPYEIWVEAT NRLGSARSDV LTLDIL
CRLF1 encodes a member of the cytokine type I receptor family. The protein forms a secreted complex with cardiotrophin-like cytokine factor 1 and acts on cells expressing ciliary neurotrophic factor receptors. The complex can promote survival of neuronal cells. Mutations in this gene result in Crisponi syndrome and cold-induced sweating syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: class I cytokine receptor; CLF-1; Cytokine receptor-like factor 1; cytokine type 1 receptor CRLP-1; Cytokine-like factor 1; ZcytoR5
Gene Aliases: CISS; CISS1; CLF; CLF-1; CRLF1; NR6; UNQ288/PRO327; zcytor5
UniProt ID: (Human) O75462
Entrez Gene ID: (Human) 9244
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support