Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibodies Inc
For maximum recovery of product, centrifuge the vial prior to removing the cap.
CACNB3 encodes a regulatory beta subunit of the voltage-dependent calcium channel. Beta subunits are composed of five domains, which contribute to the regulation of surface expression and gating of calcium channels and may also play a role in the regulation of transcription factors and calcium transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAB3; Calcium channel subunit beta 3; Calcium channel voltage-dependent subunit beta 3; calcium channel, voltage-dependent, beta 3 subunit; Cav; CCHB3; Voltage-dependent L-type calcium channel subunit beta-3
Gene Aliases: Beta3; Ca(v)beta3; CAB3; CACH3B; CACNB3; CACNLB3; Cchb3
UniProt ID: (Human) P54284, (Rat) P54287, (Mouse) P54285
Entrez Gene ID: (Human) 784, (Rat) 25297, (Mouse) 12297
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support