Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5833447 Nurture your progress Savings to support your research. Order before 27 June . 詳細へ
FIGURE: 1 / 3
This antibody is predicted to react with Monkey, Mouse, Rat.
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
DENND5A (DENN domain-containing protein 5A) is a guanine nucleotide exchange factor (GEF) which activates RAB6A and RAB39A/RAB39B. It promotes the exchange of GDP to GTP by converting inactive GDP-bound Rab proteins to their active GTP form. DENND5A is also involved in the negative regulation of neurtie outgrowth. Mutations affecting this gene can result in epileptic encephalopathy, early infantile, 49 (EIEE49).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DENN domain-containing protein 5A; DENN/MADD domain containing 5A; RAB6 interacting protein 1; Rab6-interacting protein 1; Rab6IP1
Gene Aliases: 1500012B19Rik; AJ245569; AW496491; DENND5A; KIAA1091; mKIAA1091; ORF37; RAB6IP1
UniProt ID: (Human) Q6IQ26, (Mouse) Q6PAL8, (Rat) G3V7Q0
Entrez Gene ID: (Human) 23258, (Mouse) 19347, (Rat) 308942
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support