Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 6
Recommended positive controls: 293T, A431, PC-12.
Predicted reactivity: Mouse (88%), Rat (89%), Pig (88%), Bovine (85%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Dihydrofolate reductase converts dihydrofolate into tetrahydrofolate, and is required for the synthesis of purines, thymidylic acid and certain amino acids. While the functional dihydrofolate reductase gene has been mapped to chromosome 5, multiple intronless processed pseudogenes or dihydrofolate reductase like genes have been identified on separate chromosomes. Dihydrofolate reductase deficiency has been linked to megaloblastic anemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dihydrofolate reductase; Dihydrofolate reductase 1 (active)
Gene Aliases: DHFR; Dhfr1; DHFRP1; DYR
UniProt ID: (Human) P00374, (Rat) Q920D2
Entrez Gene ID: (Human) 1719, (Rat) 24312
Molecular Function:
reductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support