Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: Mouse testis.
Immunogen sequence: MSRPGHGGLM PVNGLGFPPQ NVARVVVWEW LNEHSRWRPY TATVCHHIEN VLKEDARGSV VLGQVDAQLV PYIIDLQSMH QFRQDTGTMR PVRRNFYDPS SAPGKGIVWE WENDGGAWTA YDMDICITIQ NAYEKQHPWL DLSSLGFCYL IYFNSMSQMN RQTRRRRRLR RRLDLAYPLT VGSIPKSQSW
Studies in Drosophila have identified this gene as encoding a positive regulator of the Notch-signaling pathway. The human gene encodes a protein of unknown function; however, it may play a role in basic helix-loop-helix transcription factor activity.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: deltex 1; deltex 1, E3 ubiquitin ligase; deltex homolog 1; Deltex1; E3 ubiquitin-protein ligase DTX1; fractionated x-irradiation induced transcript 1; FXI-T1; hDTX1; mDTX1; Protein deltex-1; RING-type E3 ubiquitin transferase DTX1
Gene Aliases: DTX1; Fxit1; hDx-1; mKIAA4160
UniProt ID: (Human) Q86Y01, (Mouse) Q61010
Entrez Gene ID: (Human) 1840, (Mouse) 14357
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support