Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MVRGRISRL SVRDVRFPTS LGGHGADAMH TDPDYSAAYV VIETDAEDGI KGCGITFTLG KGTEVVVCAV NALAHHVLNK DLKDIVGDFR GFYRQLTSDG QLRWIGPEKG VVHLATAAVL NAVWDLWAKQ EGKPVWKLLV DMDPRMLVSC IDFRYITDVL TEEDALEILQ KGQIGKKERE KQMLAQGYPA YTTSCAWLGY SDDTLKQLCA QALKDGWTRF KVKVGADLQD DMRRCQIIRD MIGPEKTLMM DANQRWDVPE AVEWMSKLAK FKPLWIEEPT SPDDILGHAT ISKALVPLGI GIATGEQCHN RVIFKQLLQA KALQFLQIDS CRLGSVNENL SVLLMAKKFE IPVCPHAGGV GLCELVQHLI IFDYISVSAS LENRVCEYVD HLHEHFKYPV MIQRASYMPP KDPGYSTEMK EESVKKHQYP DGEVWKKLLP AQEN
DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. Human proteins MECP2, MBD1, MBD2, MBD3, and MBD4 comprise a family of nuclear proteins related by the presence in each of a methyl-CpG binding domain (MBD). Each of these proteins, with the exception of MBD3, is capable of binding specifically to methylated DNA. MECP2, MBD1 and MBD2 can also repress transcription from methylated gene promoters. In contrast to other MBD family members, MECP2 is X-linked and subject to X inactivation. MECP2 is dispensible in stem cells, but is essential for embryonic development. MECP2 gene mutations are the cause of most cases of Rett syndrome, a progressive neurologic developmental disorder and one of the most common causes of mental retardation in females.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antisense RNA to thymidylate synthase; HSRTSBETA; L-fuconate dehydratase; Mitochondrial enolase superfamily member 1; rTS; rTS alpha; rTS beta
Gene Aliases: ENOSF1; RTS; TYMSAS
UniProt ID: (Human) Q7L5Y1
Entrez Gene ID: (Human) 55556
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support