Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: HDHPRPESK SETEARRSAI KRQMASFYQP QKKRIRESEA EENQDSSGHF SNIPPLENPE DFDIVTETSF PIPGQPCPSF PKSDVYKATC DLATFQGDKM PPFQKYSSPR IYLPRPPCSY ELANPGYTNS SPYPTLYKDS TSIPNDTDWV HLNTLQCNVN SYSSYERSFD FTNKQHGWKP ALGKPSLVER TNHGQFQAMA TRPYYNPELP CRYLTTPPPG APALQTVITT TTKVSYQAYQ PPAMKYSDSV REVKSLSSCN YAPEDTGMSV YPEPWGPPVT VTRAASPSGP PPMKIAGDCR AIRPTVAIPH EPVSSRTDEA ETWDVCLSGL GSAVSYSDRV GPFFTYNNED F (157-506 aa encoded by BC069603)
This gene is a homolog of the Drosophila glial cells missing gene, which is thought to act as a binary switch between neuronal and glial cell determination. The protein encoded by this gene contains a conserved N-terminal GCM motif that has DNA-binding activity. The protein is a transcription factor that acts as a master regulator of parathyroid development. It has been suggested that this transcription factor might mediate the effect of calcium on parathyroid hormone expression and secretion in parathyroid cells. Mutations in this gene are associated with hypoparathyroidism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chorion-specific transcription factor GCMb; GCM motif protein 2; glial cells missing 2; Glial cells missing homolog 2; glial cells missing homolog b; glide/gcm protein homolog; hGCMb; mGCMb; RP3-417M14.3
Gene Aliases: Gcm-rs1; Gcm1; Gcm1-rs2; GCM2; GCMB; hGCMb
UniProt ID: (Human) O75603, (Mouse) O09102
Entrez Gene ID: (Human) 9247, (Mouse) 107889
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support