Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Positive Control - WB: human HepG2 whole cell, human THP-1 whole cell, human U251 whole cell, monkey COS-7 whole cell, rat C6 whole cell, mouse NIH/3T3 whole cell. IHC: Human Lung Cancer tissue, Human Intestinal Cancer tissue, Rat Cardiac Muscle tissue. ICC/IF: U20S cell, NIH3T3 Cell. Flow: U87 cell.
The protein encoded by this gene binds to the "plus" ends of actin monomers and filaments to prevent monomer exchange. The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments. Defects in this gene are a cause of familial amyloidosis Finnish type (FAF). Multiple transcript variants encoding several different isoforms have been found for this gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Actin-depolymerizing factor; ADF; ADFBrevin; AGEL; Brevin; DKFZp313L0718; Gelsolin; RP11-477J21.1
Gene Aliases: ADF; AGEL; Gsb; GSN
UniProt ID: (Human) P06396, (Rat) Q68FP1, (Mouse) P13020
Entrez Gene ID: (Human) 2934, (Rat) 296654, (Mouse) 227753
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support