Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAARSVSGIT RRVFMWTVSG TPCREFWSRF RKEKEPVVVE TVEEKKEPIL VCPPLRSRAY TPPEDLQSRL ESYVKEVFGS SLPSNWQDIS LEDSRLKFNL LAHLADDLGH VVPNSRLHQM CRVRDVLDFY NVPIQDRSKF DELSASNLPP NLKITWSY
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a putative 39S subunit protein and belongs to the L47P ribosomal protein family. Pseudogenes corresponding to this gene are found on chromosomes 2p, 2q, 5p, and 10q. [provided by RefSeq, Jul 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 39S ribosomal protein L50, mitochondrial; L50mt; Large ribosomal subunit protein mL50; mitochondrial 39S ribosomal protein L50
Gene Aliases: MRP-L50; MRPL50
UniProt ID: (Human) Q8N5N7
Entrez Gene ID: (Human) 54534
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support