Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-31748 has been tested in indirect ELISA.
NMDA receptor subtype of glutamate-gated ion channels with reduced single-channel conductance, low calcium permeability and low voltage-dependent sensitivity to magnesium. Mediated by glycine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GluN3B; glutamate [NMDA] receptor subunit 3B; glutamate receptor; Glutamate receptor ionotropic, NMDA 3B; glutamate receptor, ionotropic, N-methyl-D-aspartate 3B; N-methyl-D-aspartate receptor subtype 3B; NMDA receptor subunit 3B; NMDA type glutamate receptor subunit NR3B; NMDAR3B
Gene Aliases: GluN3B; GRIN3B; NR3B
UniProt ID: (Human) O60391
Entrez Gene ID: (Human) 116444
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support