Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
FIGURE: 1 / 4
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
After binding acetylcholine, the ACHR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetylcholine receptor; acetylcholine receptor, nicotinic, alpha 2 (neuronal); cholinergic receptor; cholinergic receptor, nicotinic alpha 2; cholinergic receptor, nicotinic, alpha 2 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 2 (neuronal); Neuronal acetylcholine receptor subunit alpha-2; nicotinic acetylcholine receptor
Gene Aliases: ACHR; Acra-2; Acra2; BC011490; CHRNA2
UniProt ID: (Human) Q15822, (Rat) O08952, (Mouse) Q91X60
Entrez Gene ID: (Human) 1135, (Rat) 170945, (Mouse) 110902
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support