Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
This antibody is predicted to react with rat based on sequence homology.
GTPase-activating protein (GAP) that stimulates the GTPase activity of Rho-type GTPases. Thereby, controls Rho-type GTPases cycling between their active GTP-bound and inactive GDP-bound states. Acts as a GAP at least for CDC42 and RAC1 (PubMed:11431473). In neurons, is involved in dendritic spine formation and synaptic plasticity in a specific RAC1-GAP activity. Limits the initiation of exploratory dendritic filopodia. Recruited to actin-patches that seed filopodia, binds specifically to plasma membrane sections that are deformed inward by acto-myosin mediated contractile forces. Acts through GAP activity on RAC1 to reduce actin polymerization necessary for filopodia formation. In association with SHANK3, promotes GRIA1 exocytosis from recycling endosomes and spine morphological changes associated to long-term potentiation. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NPC-A-10; Rho GTPase-activating protein 44; rho GTPase-activating protein RICH2; Rho-type GTPase-activating protein RICH2; RhoGAP interacting with CIP4 homologs protein 2; RICH-2
Gene Aliases: ARHGAP44; KIAA0672; NPC-A-10; RICH2
UniProt ID: (Human) Q17R89
Entrez Gene ID: (Human) 9912
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support