Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: HPLIPTDYIK NLPMWRFKCL GVQSEEEMSE GSQDTENDSE SENTDTLSSE KEVFEDKQSV LHNSPGTASH CDAEACSCAN KYCQTSPCER KGRSYGSYN
The protein encoded by this gene contains a RING-H2 finger, a motif known to be involved in protein-protein and protein-DNA interactions. This gene is highly expressed in normal cerebellum, but not in the cerebral cortex. The expression of the rat counterpart in the frontal cortex and hippocampus was shown to be induced by elctroconvulsive treatment (ECT) as well as chronic antidepressant treatment, suggesting that this gene may be a molecular target for ECT and antidepressants. [provided by RefSeq].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase RNF103; hKF-1; KF-1; RING finger protein 103; RING-type E3 ubiquitin transferase RNF103; Zfp-103; Zinc finger protein 103 homolog; zinc finger protein expressed in cerebellum
Gene Aliases: HKF-1; KF-1; KF1; RNF103; ZFP-103; ZFP103
UniProt ID: (Human) O00237
Entrez Gene ID: (Human) 7844
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support