Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: MCF-7.
Predicted reactivity: Human (99%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Spermatogenesis associated 6-like protein
Gene Aliases: bA6J24.2; C9orf68; SPATA6L
UniProt ID: (Human) Q8N4H0
Entrez Gene ID: (Human) 55064
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support