Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 11
Recommended positive controls: 293T, HeLa, rat brain, mouse brain.
Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (81%), Xenopus laevis (83%), Chicken (90%), Bovine (99%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
STRAP is a 350 amino acid MAP activator with WD-40 repeats that is localized both in the cytoplasm and the nucleus. It is a ubiquitously expressed protein that acts as an inhibitor of TGF-beta signaling and an important regulator of cell proliferation. Stable expression of STRAP results in activation of the mitogen-activated protein kinase/extracellular signal-regulated kinase pathway and in down-regulation of the cyclin-dependent kinase inhibitor p21 (Cip1), which results in retinoblastoma protein hyperphosphorylation. Upregulation of STRAP might play a role in tumor development as an early event for colorectal cancers. It is the first component of the U snRNP assembly machinery that associates with SMN complex in a compartment-specific way and plays a crucial role in the intracellular distribution of SMN1.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MAP activator with WD repeats; Serine-threonine kinase receptor-associated protein; UNR-interacting protein; WD-40 repeat protein PT-WD
Gene Aliases: AW557906; C78091; C79202; MAWD; PT-WD; STRAP; UNRIP
UniProt ID: (Human) Q9Y3F4, (Mouse) Q9Z1Z2, (Rat) Q5XIG8
Entrez Gene ID: (Human) 11171, (Mouse) 20901, (Rat) 297699
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support