Osmium • Transition Metal
Primary XPS region: Os4f
Overlapping regions: Zr4s, Fe3p, Ir3p, Li1s, Pt5p3
Binding energies of common chemical states:
Chemical stateBinding energy Os4f7/2
Os metal52 eV
Experimental information
N/A
Interpretation of XPS spectra
N/A
General comments
  • Os4d peaks overlap with C1s region, necessitating careful peak fitting.
About this element
Element Crystal hex
Symbol: Os
Date of discovery: 1803
Name origin: Greek osmë
Appearance: blue-white
Discoverer: Smithson Tenant
Obtained from: platinum ores
Melting point: 3306 K
Boiling point: 5285 K
Density[kg/m3]: 22610
Molar volume: 8.42 × 10-6 m3/mol
Protons/Electrons: 76
Neutrons: 114
Shell structure: 2,8,18,32,14,2
Electron configuration: [Xe]4f145d66s2
Oxidation state: 3,4
Crystal structure: hexagonal
Discovered by S. Tenant in 1803 from the residue of crude platinum, this lustrous, blue-white metallic element is extremely hard and brittle, even at high temperatures. The densest of all elements, osmium has the highest melting point and lowest vapor pressure of the platinum group. While the metal form is often difficult to isolate, osmium powder can be sintered at high temperatures. This powder gives off a highly toxic, strong-smelling tetraoxide, giving osmium its Greek origin namesake "osme", meaning odor.
Products
Nexsa G2 XPS
  • Micro-focus X-ray sources
  • Unique multi-technique options
  • Dual-mode ion source for monoatomic & cluster ion depth profiling
Style Sheet for Instrument Cards Original
K-Alpha XPS
  • High resolution XPS
  • Fast, efficient, automated workflow
  • Ion source for depth profiling
ESCALAB QXi XPS
  • High spectral resolution
  • Multi-technique surface analysis
  • Extensive sample preparation and expansion options
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.