Confidently process complex samples with chemistry that is up for the challenge
The PrepFiler and PrepFiler BTA Forensic DNA Extraction Kits are part of an integrated solution that includes proven reagents for use for use in manual, semi-automated and high-throughput workflows to provide reliable, simplified, and time-saving DNA extraction and purification.
The kits use magnetic particles with an optimized multi-component surface chemistry to deliver robust and reliable DNA yield from a wide range of forensic sample types.
Tested sample types
Kit
Sample types
PrepFiler Forensic DNA Extraction kits
Routing forensic sample types, including:
  • Body fluids (blood, saliva, semen)
  • Stains and swabs of body fluids
  • Hair roots
  • Touch/trace DNA samples
PrepFiler BTA Forensic DNA Extraction kits
The most challenging forensic sample types, including:
  • Bone
  • Teeth
  • Adhesive substrates (chewing gum, cigarette butts, tape lifts)
  • Touch/trace DNA samples
Featured videos
Enhancing Workflow Efficiency—DNA Labs International's Integration of Innovative Technologies
DNA Labs International has successfully leveraged cutting-edge technologies, including the HID NIMBUS Presto System, to optimize their workflow efficiency and increase productivity.
Watch video ›
How Efficient is Your DNA Extraction Method? A Multi-year, Multi-lab Review of GEDNAP Proficiency Test Results
A presentation of the German DNA Profiling Group (GEDNAP) multi-year, multi-lab, extraction proficiency tests and insights into various DNA extraction methods.
Watch video ›
Key features of both kits
  • Improves the overall yield, concentration, and purity of DNA isolated from forensic samples, while enabling removal of PCR inhibitors
  • Improves the downstream genotyping success rate, to help ensure that critical and often limited casework samples can be successfully analyzed
  • Contains specially developed magnetic particles and reagents that optimize DNA binding and elution of DNA in a small volume
  • Offers streamlined protocols to process an extensive variety of forensic sample types
  • The PrepFiler BTA kit includes PrepFiler BTA Lysis Buffer to successfully process more challenging samples
prepfiler-dna-extraction
 Click image to enlarge
Manual, automated, and express formats
There are three kit format options, for both PrepFiler and PrepFiler BTA, to fit all workflows:
Kit
Workflow
PrepFiler Forensic DNA Extraction Kit
For manual pipetting
PrepFiler BTA Forensic DNA Extraction Kit
PrepFiler Automated Forensic DNA Extraction Kit
For use with any automated liquid handler
PrepFiler BTA Automated Forensic DNA Extraction Kit
PrepFiler Express Forensic DNA Extraction Kit
For use with the AutoMate Express instrument
PrepFiler Express BTA Forensic DNA Extraction Kit
applied-biosystems-family-products-
Ordering information
For Research, Forensic, or Paternity Use Only. Not for use in diagnostic procedures.
Style Sheet for Global Design System