Cell death can occur via two processes–Accidental Cell Death (ACD) or Programmed Cell Death (PCD). Apoptosis is a type of PCD that is essential for maintaining the homeostasis of the cell and doubles as a defense mechanism in pathological conditions. Though first described by Carl Vogt in 1842, the term apoptosis was first used in 1972 by John Foxton Ross Kerr while distinguishing this specific mechanism from necrosis. While apoptosis is highly regulated, necrosis is cell death by injury and is not directly controlled by the cell. This article offers a brief overview of the major apoptotic mechanisms and key players along the various pathways.
Diagram of apoptosis versus necrosis
Figure 1. Comparison of apoptosis and necrosis.
Morphological and biochemical changes associated with apoptosis
The hallmarks of apoptosis can be classified into changes pertaining to the nucleus, cell membrane and cytosolic components, and mitochondria function. Chromatin condensation (pyknosis), as well as nuclear and DNA fragmentation (karyorrhexis), are the significant nuclear changes associated with apoptosis. Following apoptosis initiation, the cellular junctions are lost, the cell membrane shrinks, and the cytosolic components are organized into apoptotic bodies. The presence of phosphatidylserine in the outer leaflet of the cell membrane marks the apoptotic bodies for phagocytic elimination.
In addition, cytoskeletal components and structural proteins like actin and spectrin are also deregulated, leading to a loss of cellular integrity. Apoptotic mechanisms can further trigger the release of pro-apoptotic factors from the mitochondria like cytochrome C, AIF, and SMAC/DIABLO.
Explore Apoptosis Assays
There are two major cellular signaling pathways that orchestrate the apoptotic processes—intrinsic and extrinsic.
Intrinsic Pathway
The apoptosis intrinsic pathway is triggered by stimuli causing DNA damage or metabolic stress. This results in the activation of pro-apoptotic BAX and BAK belonging to the Bcl2 family of proteins. BAX/BAK activation leads to mitochondrial outer membrane permeabilization (MOMP) and the release of cytochrome C, CYCS, SMAC/DIABLO, and HTRA2 into the cytoplasm. Cytochrome-C associates with cytoplasmic APAF1, leading to the formation of the apoptosome complex, which activates caspase-9. Active caspase-9, in turn, cleaves and activates the executioner caspases (caspases-3, 6, 7) that help in DNA fragmentation and the ensuing morphological and biochemical changes in apoptosis. Below are western blot and immunofluorescent data examples looking at two intrinsic targets: cytochrome C and DIABLO.
Western blot analysis of cytochrome c1 in various cell lines
Figure 2. Cytochrome C1 antibody in western blot. Western blot analysis was performed on whole cell extracts (30 µg lysate) of HeLa (Lane 1), A-431 (Lane 2), Hep G2 (Lane 3), MCF7 (Lane 4), HT-29 (Lane 5), K-562 (Lane 6) and MDA-MB-231 (Lane 7). The blot was probed with Cytochrome C1 Polyclonal Antibody (Cat. No. PA5-27490, 1 µg/mL) and detected by chemiluminescence using Goat anti-Rabbit IgG (H+L) Superclonal Recombinant Secondary Antibody, HRP (Cat. No. A27036, 0.25 µg/mL, 1:4,000 dilution). A 34 kDa band corresponding to Cytochrome C1 was detected across the cell lines tested.
Figure 3. DIABLO antibody in Immunofluorescence. Immunofluorescent analysis of DIABLO (green) showing staining in the cytoplasm of MCF-7 cells. Formalin-fixed cells were permeabilized with 0.1% Triton X-100 in TBS for 5-10 minutes and blocked with 3% BSA-PBS for 30 minutes at room temperature. Cells were probed with a DIABLO monoclonal antibody (SMAC 17 1-87) (Cat. No. MA1-936) in 3% BSA-PBS at a dilution of 1:100 and incubated overnight at 4°C in a humidified chamber. Cells were washed with PBST and incubated with a DyLight-conjugated secondary antibody in PBS at room temperature in the dark. F-actin (red) was stained with a fluorescent red phalloidin and nuclei (blue) were stained with Hoechst or DAPI. Images were taken at a magnification of 60x.
Immunofluorescent analysis of DIABLO in MCF-7 cells
Extrinsic Pathway
The apoptosis extrinsic pathway is triggered by diverse stimuli or the withdrawal of stimuli (e.g. growth factors). Apoptosis triggered by the extrinsic pathway involves transmembrane receptor interactions with respective ligands. These ‘death receptors’ belonging to the TNF gene superfamily consist of cysteine-rich extracellular domains and conserved intracellular ‘death domains’. The best studied ligand-receptor pairs are FasL/FasR, TNFα/TNFR1, Apo3L/DR3, Apo2L/DR4/DR5, and TRAIL/TRAILR1. Ligand binding to FasR and TNFR1 induces receptor clustering and recruitment of cytoplasmic adaptors FADD and TRADD, respectively. This results in the formation of DISC (death-inducing signaling complex) that further cleaves pro-caspase-8 to generate active caspase 8. Caspase 8 induces cleavage and myristoylation of cytoplasmic BID, a pro-apoptotic Bcl-2 family protein, and induces its translocation into mitochondria while it also activates the executioner caspases directly. Thus, BID acts as a common player between intrinsic and extrinsic apoptotic pathways. Below are western blot and immunofluorescent data examples looking at two extrinsic pathway targets: DR3 and BID.
Western blot analysis of DR3 in various whole cell extracts and tissue lysates
Figure 4. DR3 antibody in western blot. Western blot analysis was performed on whole cell extracts (30 µg lysate) of K-562 (Lane 1), Jurkat (Lane 2), MOLT4 (Lane 3), THP-1 (Lane 4) and tissue lysates of (30 µg lysate) Mouse Thymus (Lane 5). The blots were probed with DR3 Recombinant Rabbit Monoclonal Antibody (11H6L9) (Cat. No. 702277, 1-2 µg/mL) and detected by chemiluminescence using Goat anti-Rabbit IgG (H+L) Superclonal Recombinant Secondary Antibody, HRP (Cat. No. A27036, 0.4 µg/mL, 1:2,500 dilution). Three isoforms at 45, 41 and, 19 kDa corresponding to DR3 was observed across cell lines and tissue tested.
Figure 5. BID antibody in immunofluorescence. Immunofluorescent analysis was performed using 70% confluent log phase A-431 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton X-100 for 15 minutes and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled with BID Recombinant Rabbit Monoclonal Antibody (JM11-14) (Cat. No. MA5-32642) at 5 µg/mL in 0.1% BSA, incubated at 4oC overnight and then labeled with Goat anti-Rabbit IgG (H+L), Superclonal Recombinant Secondary Antibody, Alexa Fluor 488 (Cat. No. A27034) at a dilution of 1:2,000 for 45 minutes at room temperature (Panel a: green). Nuclei (Panel b: blue) were stained with ProLong Diamond Antifade Mountant with DAPI (Cat. No. P36962). F-actin (Panel c: red) was stained with Rhodamine Phalloidin (Cat. No. R415). Panel d represents the merged image showing cytoplasmic localization. Panel e represents control cells with no primary antibody to assess background. The images were captured at 60X magnification.
Key apoptosis targets
Apoptosis pathways have multiple steps and multiple targets that need to be available for the process to proceed as outlined above. Here are some key targets to consider when researching apoptosis.
Caspases
The studies on the CED-3 gene in nematode C. elegans led to the discovery of a homologous family of proteins in mammals, called caspases. These are cysteine aspartic-specific proteases that orchestrate the apoptotic processes. They are expressed as inactive zymogens in the cell and are activated via cleavage of pro-domains and inter-subunit linkers. The 14 different caspases in mammals are broadly classified into 3 groups based on sequence similarities and biological functions:
  • Initiators (Caspases 2, 8, 9, 10) possess long N-terminal caspase recruitment domains (CARD). Of these, Caspase-2 is instrumental in apoptosis and is triggered by DNA damage, metabolic deregulations, and endoplasmic reticulum stress
  • Executioners (Caspases 3, 6, 7, 14) are characterized by short pro-domains called death effector domains.
  • Inflammatory (Caspases 1, 4, 5, 11, 12, 13) caspases possess long caspase recruitment domains (CARD) like initiator caspases, but prefer aromatic or hydrophobic residues in the P4 position of substrates while initiator caspases prefer leucine or valine residues.
Bcl-2 family of proteins
Bcl-2 family proteins are classified based on their pro- or anti-apoptotic action and the Bcl-2 Homology (BH) domains. Anti-apoptotic/pro-survival Bcl-2-like proteins (e.g. Bcl-2, Bcl-xL, Bcl-w, Mcl-1, and A1/Bfl-1) and pro-apoptotic/anti-survival BAX-like proteins (e.g. BAX, BAK, and BOK/Mtd) display 4 BH domains. In contrast, the pro-apoptotic BH3-only proteins (e.g. BID, Bim/Bod, BAD, Bmf, BIK/Nbk, BLK, NOXA, PUMA/Bbc3, and HRK/DP5) have only a short BH3 domain. The BH3 only proteins are activated upon various stress signals and in turn inactivate the pro-survival Bcl-2-like proteins. Subsequently, the pro-apoptotic proteins like BAX and BAK initiate the mitochondrial pathway leading to apoptosis. Below is a western blot analysis that confirms antibody specificity of a Bcl-2 antibody that would be suitable for apoptosis research and a flow cytometry analysis of Mcl-1 in human peripheral blood cells.
Western blot analysis of Bcl-2 in various cell lines
Figure 6. (A) Bcl-2 antibody in western blot. Western blot analysis was performed on whole cell extracts (30 µg lysate) of MCF-7 (Lane 1), Raji (Lane 2), T-47D (Lane 3), MDA-MB-231 (Lane 4), HeLa (Lane 5), and Jurkat (Lane 6). The blot was probed with Bcl-2 Monoclonal Antibody (124) (Cat. No. MA1-26233, 1:500 dilution) and detected by chemiluminescence using Goat anti-Mouse IgG (H+L) Superclonal Recombinant Secondary Antibody, HRP (Cat. No. A28177, 0.25 µg/mL, 1:4,000 dilution). A 27 kDa band corresponding to Bcl-2 was observed across the cell lines tested. (B) Knockdown of Bcl-2 was achieved by transfecting HeLa cells with Bcl-2 specific validated siRNAs. Western blot analysis (A) was performed using whole cell extracts from the Bcl-2 knockdown cells (lane 3), non-specific scrambled siRNA transfected cells (lane 2) and untransfected cells (lane 1). The blots were probed with the same Bcl-2 Monoclonal Antibody at a 1:500 dilution) and the Goat anti-Mouse IgG (H+L) Superclonal Secondary Antibody, HRP at a 1:4,000 dilution. Densitometric analysis of this western blot is shown in histogram (B). Decrease in signal upon siRNA mediated knock down confirms that antibody is specific to Bcl-2.
Figure 7. Phospho-Mcl-1 (SER159) antibody in flow. Top: Intracellular staining of normal human peripheral blood cells that were untreated (left) or treated with Calyculin A for 4 hours (right) with CD3 Monoclonal Antibody (SK7), PerCP-Cyanine5.5 (Cat. No. 45-0036-42) and phospho-Mcl-1 (S159) PE. Plots show cells in the lymphocyte gate. Bottom: Normal human peripheral blood cells were unstimulated (orange histogram), were stimulated with CD3 Monoclonal Antibody (OKT3), Functional Grade (Cat. No. 16-0037-81) and CD28 Monoclonal Antibody (CD28.2), Functional Grade (Cat. No. 16-0289-81) in the presence of the proteasome inhibitor MG-132 (purple histogram) or were treated with Calyculin A (green histogram). The cells were then intracellularly stained with CD3 Monoclonal Antibody (SK7) PerCP-Cyanine5.5 (Cat. No. 45-0036-42) and Phospho-Mcl-1 (Ser159) Monoclonal Antibody (RBCERNR), PE (Cat. No. 12-9038-42) using the Intracellular Fixation and Permeabilization Buffer Set (Cat. No. 88-8824-00) and protocol. CD3+ cells in the lymphocyte gate were used for analysis.
IAPs
The inhibitor of apoptosis proteins (IAPs) were first identified in insect baculoviruses. Cellular and viral IAPs are characterized by the presence of baculoviral repeat domain (BIR) repeats. The most widely studied mammalian IAPs are survivin, cellular IAP1 (cIAP1), cIAP2, and X-linked IAP (XIAP). Among the IAPs, only XIAP can directly bind and inhibit caspases (caspases-3 and -9). Upon apoptotic stimuli, IAP inhibitors, including SMAC/DIABLO and HtrA2/Omi, are released from the mitochondria and bind to XIAP’s BIR domains, releasing active caspases into the cytosol. In contrast, cIAP1and cIAP2 bind to tumor necrosis factor (TNF) receptor-associated factors (TRAFs) and inhibit cell death induced by TNF receptor 1 (TNFR1) by triggering the pro-survival signaling pathways. Below is immunofluorescent analysis of surviving in the cell cycle stages of HeLa cells.
Survivin
Composite
Survivin
Composite
Immunofluorescent analysis of survivin in the cell cycle stages of HeLa cells
Immunofluorescent analysis of survivin in the cell cycle stages of HeLa cells
Figure 8. Survivin antibody in IF. Immunofluorescence analysis of survivin using Survivin Monoclonal Antibody (1H5) (Cat. No. MA5-17035) shows differential subcellular localization of Survivin at various cell cycle stages in HeLa cells- low levels at early prophase and an increase at late prophase. During metaphase survivin is localized in the centromeres, remains in the spindle midzone at anaphase and localizes in the midbody between two daughter cells at telophase.
TNF superfamily
The TNF superfamily consists of the death receptors (DRs) and their ligands such as TNF, TRAIL, and Fas ligand (FasL). The binding of these ligands to their respective receptors trigger extrinsic apoptosis pathways. CD95 (DR2/Fas/APO-1), TNF receptor 1 (DR1/TNFR1), TRAIL-R1 (DR4), and TRAIL-R2 (DR5) are the well characterized DRs that bind CD95 ligand (CD95L/FasL), TNFα, lymphotoxin-α (both of these bind to TNFRI), and TRAIL (binds to TRAIL-R1 and TRAIL-R2), respectively.
The interaction of Fas (CD95/APO-1/DR2) with FasL results in conformational changes in the death domain (DD) of the receptor, which recruits FADD through its DD. FADD, in turn, interacts with pro-caspase-8 and -10 and activates them via auto-cleavage, thus triggering the downstream apoptotic signaling pathways. The CD95/FADD/caspase-8/-10 complex is termed the ‘death-inducing signaling complex’ (DISC).
Similarly, TRAIL binds to its receptors, TRAIL receptor 1 (death receptor 4, DR4) and TRAIL receptor 2 (death receptor 5, DR5) and recruits the adaptor protein FADD along with inactive caspase-8 and -10 to form the DISC complex. The cleaved caspase-8 and -10 are then released into the cytosol to activate effector caspases.
TNF-mediated apoptosis is generally carried out by TNFR1 (DR1). Binding of TNF to TNFR1 triggers the recruitment of TRADD through its DD. TRADD interacts with FADD resulting in the recruitment of pro-caspase-8, which is proteolytically cleaved to active caspase-8. Caspase-8, in turn, activates caspase-3. 
Cellular FLICE (FADD-like IL-1β-converting enzyme)-inhibitory protein (c-FLIP) is a master regulator that inhibits TNF-α, Fas-L and TRAIL-induced apoptosis. c-FLIP binds to FADD and/or caspase-8 or -10 and DR5 and forms an apoptosis inhibitory complex (AIC), which prevents DISC formation.
p53
p53 is a transcription factor that binds to DNA in a sequence-specific manner. p53 can regulate both intrinsic and extrinsic pathways of apoptosis by inducing transcription of several proteins like CD95, TRAILR2, PUMA, BID and BAX. p53 also translocates to the mitochondria in response to apoptotic signals where it forms inhibitory complexes with Bcl-XL and Bcl-2 causing the permeabilization of the mitochondrial membrane and cytochrome c release. Moreover, cytosolic p53 might induce the activation of pro-apoptotic BAX via direct protein-protein interactions.
Emerging cell death pathways
Various novel non-apoptotic forms of regulated cell death have also emerged and are increasingly being implicated in multiple human diseases. Necroptosis is one such mechanism that works against pathogen-mediated infections. It is characterized by cell swelling followed by plasma membrane rupture and chromatin condensation. Generally, necroptosis is initiated upon caspase-8 inhibition and is mediated by ligands including FasL, TNF, and LPS leading to activation of RIPK3 (RIP3). The formation of necrosomes with RIPK1/RIPK3 and mixed lineage kinase domain-like pseudo kinase (MLKL) further leads to MLKL activation and its trans-location to the plasma membrane to initiate cell death. The figure below shows the validation of RIP3 antibody using relative expression approach. Other emerging cell death pathways include pyroptosis, ferroptosis, parthanatos, autophagy, and NETotic cell death that can be triggered by oxidative stress.
Figure 9. RIP3 antibody in ICC. HT-29 cells were fixed and permeabilized for detection of endogenous RIP3 using RIP3 Recombinant Polyclonal Antibody (Cat. No 711691, 2 µg/mL) and labeled with Goat anti-Rabbit IgG (H+L) Superclonal Recombinant Secondary Antibody, Alexa Fluor 488 (Cat. No. A27034, 1:2000). (A) shows representative cells that were stained for detection and localization of RIP3 protein (green), (B) is stained for nuclei (blue) using SlowFade Gold Antifade Mountant with DAPI (Cat. No. S36938). (C) represents cytoskeletal F-actin staining using Rhodamine Phalloidin (Cat. No. R415, 1:300). (D) is a composite image of Panels A, B and C clearly demonstrating cytoplasmic localization of RIP3. (E) represents control cells with no primary antibody to assess background. (F) represents a negative control on HCT116 cells which are reported RIP3 negative. Images were captured at 60X magnification.
Additional resources
Recommended reading
  1. Tang, D., Kang, R., Berghe, T.V. et al. The molecular machinery of regulated cell death. Cell Res 29, 347–364 (2019).
  2. Carneiro, B.A., El-Deiry, W.S. Targeting apoptosis in cancer therapy. Nat Rev Clin Oncol (2020).
  3. Lalaoui N, Vaux DL. Recent advances in understanding inhibitor of apoptosis proteins. F1000Res. 2018;7: F1000 Faculty Rev-1889.
  4. Kiraz, Y., Adan, A., Kartal Yandim, M. et al. Major apoptotic mechanisms and genes involved in apoptosis. Tumor Biol. 37, 8471–8486 (2016).
  5. Julien, O., Wells, J. Caspases and their substrates. Cell Death Differ 24, 1380–1389 (2017).
  6. Safa AR. c-FLIP, a master anti-apoptotic regulator. Exp Oncol. 2012;34(3):176‐184.
  7. Martinou, Jean-Claude, and Richard J Youle. Mitochondria in apoptosis: Bcl-2 family members and mitochondrial dynamics. Developmental cell vol. 21,1 (2011): 92-101.
  8. Elmore, Susan. Apoptosis: a review of programmed cell death.Toxicologic pathology vol. 35,4 (2007): 495-516.
  9. Robert Gerl, David L. Vaux. Apoptosis in the development and treatment of cancer, Carcinogenesis, Volume 26, Issue 2, February 2005, 263–270.
For Research Use Only. Not for use in diagnostic procedures.