Search Thermo Fisher Scientific
Choose from a range of environmental chambers based on the temperature, lighting and humidity ranges that your application requires. We’ve made it easy to find the right chamber for your lab.
Choosing the right Environmental Chamber depends primarily on what factors you need to control, whether temperature, humidity, lighting, CO2. Our chambers are organized around these key functions, with additional modular components and options to support your specific testing requirements. No matter which model you choose, all include rich standard features that make them easy to use and are designed to help you achieve consistently reliable results.
Heratherm Light |
Heratherm Stability Chambers |
Heratherm CO2 |
Heratherm Incubation Chambers |
Environmental control for regulated and sensitive products that require light stability testing |
Test shelf-life, stability, and other quality control demands |
Precisely control the parameters of your cell growth conditions |
Dependable microbiological incubation or temperature dependent storage and testing |
Control capability | |||
Temperature, humidity, light |
Temperature, humidity |
Temperature, humidity, CO2 |
Temperature (cooling and heating) |
Regulatory and compliance standards | |||
Enables compliance with 21 CFR Part 11. Meets ICH guidelines for photo stability testing Q1B. Simulate gradual transitions from day to night, suitable for plant and insect growth |
Enables compliance with 21 CFR Part 11. Meets compliance standards for ICH, FDA, TAPPI, ASTM or national testing standards |
Enables compliance with 21 CFR Part 11. Specific focused on temperature-only segments of drug stability and culture |
Enables compliance with 21 CFR Part 11. Specific focus on temperature-only segments of drug stability and incubation |
Size | |||
750 L (26.5 cu. ft.) |
750 L (26.5 cu. ft.) |
750 L (26.5 cu. ft.) |
750 L (26.5 cu. ft.) |
Temperature range | |||
0-70 °C 10-45 °C with light on |
0-70 °C |
0-45 °C |
0-70 °C |
Relative humidity | |||
10-90% RH |
10-90% RH |
10-90% RH |
-- |
Light | |||
LED technology |
-- |
-- |
-- |
CO2 | |||
-- |
-- |
0 - 20% v/v |
-- |
Applications | |||
ICH stability testing with UV light Plant growth Animal hatching Light simulation testing |
Stability testing Shelf-life testing Environmental testing |
Cell culture |
Precise temperature conditions above and below ambient. Excellent for microbiological incubation or storage (cooled or heated) |
Models |
|
|
|
Available for pre-order. |
Available for pre-order. |
|
Stability Chambers |
![]() Vertical Light Stability Chambers |
![]() Heat Only Reach-in Chambers |
Refrigerated Reach-in Chambers |
![]() Vertical light Refrigerated Reach-in Chambers |
Model Number |
|||||
Control Capability |
Temperature Humidity |
Temperature Humidity Light |
Elevated temperature |
Temperature |
Temperature Light |
Typical Applications |
Stability testing Shelf-life testing Product and packaging testing Materials testing Environmental stability testing Photostability testing Insect hatching Plant growth Controlled storage Freeze/thaw testing Cell culture/growth |
Microbiological incubation Insect hatching Plant growth Controlled Storage Shelf-life testing Materials testing Stability testing that does not require humidity control |
|||
Description |
Meets compliance standards for ICH, FDA, TAPPI, ASTM or national testing standards |
ICH light module for optimal light exposure according to guideline Q1B. Light modules for plant and insect growth also available |
Focused on elevated temperature-only segments of drug stability and culture |
Focused on temperature-only segments of drug stability and incubation |
Meet ICH guidelines for photo stability testing Q1B. Simulate gradual transitions from day to night, suitable for plant and insect growth |
Temperature Range |
0°C to 60°C |
10°C to 50°C with light on |
5°C above ambient to 60°C |
0°C to 60°C |
10°C to 50°C with light on |
Relative Humidity Range |
Above ambient to 95% @ 37°C (98.6°F) |
Above ambient to 75% @ 50°C (122°F) |
NA |
NA |
NA |
Lighting Modules (max.) |
2 |
2 or 3 |
NA |
NA |
3 |
Size |
11 or 29 cu. ft. |
11 or 29 cu. ft. |
29 cu. ft. |
29 cu. ft. |
29 cu. ft. |
CO2 |
Optional |
NA |
NA |
Optional |
NA |