Front angle on the MiniAmp Thermal Cycler, including the user interface home screen
Backed by the proven reliability of Applied Biosystems technology, the MiniAmp Plus Thermal Cycler and the MiniAmp Thermal Cycler were designed for routine PCR. With their compact design—just 7.5 inches (19 cm) wide—and entry-level price, MiniAmp Thermal Cyclers are small enough to maximize lab bench space.
Request quote Explore thermal cyclers
Compact thermal cycler with different block options to fit flexibility needs
Both MiniAmp Thermal Cyclers are small thermocyclers. However, the primary difference between the MiniAmp Plus Thermal Cycler and the MiniAmp Thermal Cycler is the type of block. The MiniAmp Thermal Cycler has an isothermal block, while the MiniAmp Plus Thermal Cycler offers an upgrade to a VeriFlex Block for PCR optimization.
MiniAmp Plus Thermal Cycler
Veriflex Block with three red-highlighted zones indicating the three temperature zones
VeriFlex Block (96-well, 0.2 mL)
Three independent temperature zones for easy PCR optimization
MiniAmp Thermal Cycler
An isothermal block with one red-highlighted zone indicating the one temperature zone
Isothermal block (96-well, 0.2 mL)
Uniform block temperature for basic PCR
Watch the unveiling video for MiniAmp Thermal Cyclers
Video Player is loading.
Current Time 0:00
Duration 0:37
Loaded: 26.59%
Stream Type LIVE
Remaining Time 0:37
1x
    • Chapters
    • descriptions off, selected
    • en (Main), selected
    Stay in control with Thermal Cycler Fleet Control Software
    Diagram of multiple thermal cyclers connected to one tablet to represent Fleet Control Software
    Manage multiple users, methods, and instruments with a single intuitive interface securely behind your firewall.
    Learn more about Thermal Cycler Fleet Control Software
    Ordering information
    High-performance PCR and qPCR plastics for optimal results
    Five multi-colored PCR tube plates
    Applied Biosystems PCR plastics have been designed and validated to work with our thermal cyclers for more than 25 years. Options for every format and throughput need are available to ensure consistent, precise results for end-point PCR applications. Quickly find the right fit and compatible plastics for your desired model and block formats by using our plastics selection guide.
    Explore online PCR and qPCR plastics selection tool
    Resources
    Thermal Cycler Software and Firmware Files
    Download the latest updates for your instrument
    Interested in private label thermal cyclers?
    Submit an inquiry
    Support
    Service
    We provide a full range of service options for all Applied Biosystems thermal cyclers.
    View service plans
    Multiplex Dynamic Temperature Verification Service
    Instrument qualification service: Have one of our engineers perform temperature qualifications at your site.
    View temperature service options
     Download MDTV flyer
    Order information
    Contact us for special pricing
    Catalog # Name Size Price (USD) Qty
    A37835 MiniAmp™ Plus Thermal Cycler, 1 instrument Each
    3,965.00
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    6,100.00
    Save 2,135.00 (35%)
    A37834 MiniAmp™ Thermal Cycler, 1 instrument Each
    2,983.50
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    4,590.00
    Save 1,606.50 (35%)
    For Research Use Only. Not for use in diagnostic procedures.