zeba desalting spin columns and plates
Thermo Scientific Zeba desalting products are widely used in various research applications that involve the separation of proteins, and other macromolecules from soluble low molecular weight substances (e.g., salts) that may adversely impact the stability of the analytes or interfere with downstream applications. Zeba Desalting products are also used in protein solution buffer exchange protocols for the generation of more appropriate buffers before subsequent applications such as western blotting, immunoprecipitation, activity assays, and mass spectrometry.
Zeba Desalting products yield exceptional desalting and protein-recovery characteristics and provide consistent performance over a wide range of sample sizes and protein concentrations. These desalting products speed up the desalting process by eliminating the time needed for samples to drop out of gravity flow columns. They offer a fast and efficient protocol for the collection and concentration of clean samples in one fraction, require no chromatography system, and permit multi-sample processing.
Choose the desalting product for your experiment ›
Benefits of Zeba desalting columns, plates, and cartridges:
  • High performance—proprietary resin provides excellent protein recovery and greater than 95% contaminant removal
  • Flexible—available in spin columns, filter spin plates and cartridges for a range of needs
  • Fast— spin columns eliminate the need for fraction screening or waiting for protein to emerge by gravity flow
Choose the right desalting product for your experiment
 zeba desalting spin columnzeba desalting spin columnzeba desalting spin plate
Zeba Spin Desalting Columns
Zeba 96-well Spin Plates
Zeba Chromatography Cartridges
Desalting methodSpin-column allows processing within 5-8 minutesChromatography
Sample volume range2 μL–10 mL20–100 μL15–250 μL
Resin Bed volume75 μL–20 mL550 μL1 mL–5 mL
Resin MWCO*7 kDa, 40 kDa7 kDa, 40 kDa7 kDa
AutomationNoYesYes
See all Zeba columns and plates ›
*Molecular Weight Cut-Off (MWCO)
7K Desalting Spin Products
Catalog # Name Size Price (USD) Qty
89877 Zeba™ Micro Spin Desalting Columns, 7K MWCO, 75 μL, 25 Columns Each
172.65
Online Exclusive
190.00
Save 17.35 (9%)
89878 Zeba™ Micro Spin Desalting Columns, 7K MWCO, 75 μL, 50 Columns Each
309.65
Online Exclusive
336.00
Save 26.35 (8%)
89879 Pierce™ Micro-Spin Columns Each
92.65
Online Exclusive
104.00
Save 11.35 (11%)
89882 Zeba™ Spin Desalting Columns, 7K MWCO, 0.5 mL, 25 Columns Each
162.65
Online Exclusive
176.00
Save 13.35 (8%)
89883 Zeba™ Spin Desalting Columns, 7K MWCO, 0.5 mL, 50 Columns Each
282.65
Online Exclusive
312.00
Save 29.35 (9%)
89889 Zeba™ Spin Desalting Columns, 7K MWCO, 2 mL, 5 Columns Each
68.65
Online Exclusive
75.25
Save 6.60 (9%)
89890 Zeba™ Spin Desalting Columns, 7K MWCO, 2 mL, 25 Columns Each
290.65
Online Exclusive
315.00
Save 24.35 (8%)
89891 Zeba™ Spin Desalting Columns, 7K MWCO, 5 mL, 5 Columns Each
82.65
Online Exclusive
91.50
Save 8.85 (10%)
89892 Zeba™ Spin Desalting Columns, 7K MWCO, 5 mL, 25 Columns Each
364.65
Online Exclusive
404.00
Save 39.35 (10%)
89893 Zeba™ Spin Desalting Columns, 7K MWCO, 10 mL, 5 Columns Each
103.65
Online Exclusive
115.00
Save 11.35 (10%)
89894 Zeba™ Spin Desalting Columns, 7K MWCO, 10 mL, 25 Columns Each
437.65
Online Exclusive
487.00
Save 49.35 (10%)
89934 Zeba™ Desalting Chromatography Cartridges, 7K MWCO, 1 mL, 5 columns x 1 mL Each
224.65
Online Exclusive
250.00
Save 25.35 (10%)
89935 Zeba™ Desalting Chromatography Cartridges, 7K MWCO, 5 mL, 1 column x 5 mL Each
269.65
Online Exclusive
293.00
Save 23.35 (8%)
89807 Zeba™ Spin Desalting Plates, 7K MWCO, 2 Plates Each
322.65
Online Exclusive
367.00
Save 44.35 (12%)
89808 Zeba™ Spin Desalting Plates, 7K MWCO, 4 Plates Each
608.65
Online Exclusive
660.00
Save 51.35 (8%)
A40002469 Zeba XL Spin Desalting Columns, 7K MWCO, 5 Columns 5 columns
223.65
Online Exclusive
243.00
Save 19.35 (8%)
A40002475 Zeba XL Spin Desalting Columns, 7K MWCO, 10 Columns 10 columns
441.65
Online Exclusive
478.00
Save 36.35 (8%)
40K Desalting Spin Products
Catalog # Name Size Price (USD) Qty
A40002471 Zeba XL Spin Desalting Columns, 40K MWCO, 5 Columns 5 columns
227.65
Online Exclusive
243.00
Save 15.35 (6%)
A40002476 Zeba XL Spin Desalting Columns, 40K MWCO, 10 Columns 10 columns
448.65
Online Exclusive
478.00
Save 29.35 (6%)
A57756 Zeba™ Micro Spin Desalting Columns, 40K MWCO, 25 Columns, 25 columns Each
174.65
Online Exclusive
186.00
Save 11.35 (6%)
A57758 Zeba™ Micro Spin Desalting Columns, 40K MWCO, 50 Columns, 50 columns Each
307.65
Online Exclusive
328.00
Save 20.35 (6%)
A57759 Zeba™ Spin Desalting Columns, 40K MWCO, 0.5 mL, 25 Columns, 25 columns Each
158.65
Online Exclusive
169.00
Save 10.35 (6%)
A57760 Zeba™ Spin Desalting Columns, 40K MWCO, 0.5 mL, 50 Columns, 50 columns Each
286.65
Online Exclusive
306.00
Save 19.35 (6%)
A57761 Zeba™ Spin Desalting Columns, 40K MWCO, 2 mL, 5 Columns, 5 columns Each
68.65
Online Exclusive
73.50
Save 4.85 (7%)
A57762 Zeba™ Spin Desalting Columns, 40K MWCO, 2 mL, 25 Columns, 25 columns Each
288.65
Online Exclusive
308.00
Save 19.35 (6%)
A57763 Zeba™ Spin Desalting Columns, 40K MWCO, 5 mL, 5 Columns, 5 columns Each
84.65
Online Exclusive
89.50
Save 4.85 (5%)
A57764 Zeba™ Spin Desalting Columns, 40K MWCO, 5 mL, 25 Columns, 25 columns Each
373.65
Online Exclusive
398.00
Save 24.35 (6%)
A57765 Zeba™ Spin Desalting Columns, 40K MWCO, 10 mL, 5 Columns, 5 columns Each
107.65
Online Exclusive
114.00
Save 6.35 (6%)
A57766 Zeba™ Spin Desalting Columns, 40K MWCO, 10 mL, 25 Columns, 25 columns Each
450.65
Online Exclusive
480.00
Save 29.35 (6%)
A57767 Zeba™ Spin Desalting Plates, 40K MWCO, 2 Plates, 2 plates Each
339.65
Online Exclusive
362.00
Save 22.35 (6%)
A57768 Zeba™ Spin Desalting Plates, 40K MWCO, 4 Plates, 4 plates Each
607.65
Online Exclusive
648.00
Save 40.35 (6%)
Sample desalting using gel filtration chromatography
Video Player is loading.
Current Time 0:00
Duration 0:44
Loaded: 0.00%
Stream Type LIVE
Remaining Time 0:44
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Zeba desalting products rely on gel filtration chromatography, where different molecular weight cut-off (MWCO) limits exclude molecules based on size. Compared with dialysis, this technology offers the advantages of speed, compatibility with organic and other solvents, and the ability to concentrate contaminating material down to a relatively small volume, which is useful for work with toxic/radioactive substances.
    Zeba Desalting Handbook
    Thermo Scientific Zeba Resin technology provides consistent performance and peace of mind. The unique, proprietary Zeba Resin enables better protein recovery, even for samples with low protein concentrations.
    Download the Zeba Desalting Handbook ›
    front cover of zeba desalting handbook
    For Research Use Only. Not for use in diagnostic procedures.