Bacterial RNA sequencing and studies of the plant transcriptome reveal sample complexity using Collibri Stranded RNA Library Prep Kits
Reveal the full complexity of a plant transcriptome or perform bacterial RNA sequencing using the Invitrogen Collibri Stranded RNA Library Prep kits for Illumina systems.
  • Suitable for bacterial RNA sequencing studies or plant transcriptome studies
  • <4.5 hours for library generation following ribosomal RNA (rRNA) depletion
  • For highest success rates, tracking dyes in each reagent change color when properly mixed
Library preparation for bacterial RNA sequencing or plant transcriptome studies is a two-step process.
Step 1. Deplete ribosomal RNA
The Invitrogen RiboMinus Isolation kits enable efficient and selective removal of ribosomal RNA (rRNA) from intact total RNA in less than two hours. Depleted RNA produced by RiboMinus kits is compatible with Collibri Library Prep kits in step two.
 library prep for bacterial RNA sequencing studies
RiboMinus Bacteria 2.0 Transcriptome Isolation Kit
sequence plant transcriptome studies
RiboMinus Plant Kit for RNA-Seq
Species compatibilityPan-prokaryote depletion probes have been shown to be compatible with over 76 species.Wheat, maize, rice, barley, soybean, sugar cane, sorghum, potato, cassava, Arabidopsis, tomato, tobacco, poplar, legume, and moss
Sample quality compatibilityRIN values from 5.0–10.0RIN values from 8.0–10.0
Hands-on time0.75 hours1 hour
Total time<2 hours2 hours
rRNA depleted16S
23S
5S
nuclear (25/26S and 17/18S)
chloroplast (23S and 16S)
mitochondrion (18S)
Probe design150 probes targeting conserved regions of rRNA across 76 pan-prokaryote speciesProbe mixture contains 6 probes. Each probe is specific for one or more of the conserved regions of the large cytosolic and chloroplast rRNA transcripts
Input range100 ng to 5 µg of total RNA1–10 µg total RNA
  Download manual Download manual
Step 2. Rapid library generation
 collibri mrna seq library prep
Collibri Stranded RNA Library Prep Kit for Illumina Systems
Hands-on time1.5 hours
Total time4.5 hours
RNA sourceRNA depleted of ribosomal RNA (rRNA)
Input range1–25 ng of mRNA
Species compatibilityAll
Ribosomal depletion moduleAvailable separately
Analysis difficultyBiologist-friendly
The Collibri Stranded RNA Library Prep kits for Illumina systems generate NGS libraries in 4.5 hours and maintain sample complexity. The kits use a unique protocol in which partial adaptors, also known as helper adapters, are ligated directly to single-stranded RNA prior to creation of cDNA. Full-length indexed adaptors are generated during PCR. Collibri Stranded RNA Library  Prep kits are compatible with all RNA samples following ribosomal depletion.
Figure 1. Illumina-compatible adapters are added directly to RNA fragments. Following enrichment for mRNA, transcripts are fragmented, and partial Illumina adapters are added directly to the RNA. Addition of adapters early in the process removes bias and loss of gene expression seen with older library prep kits.
Technical Note: How strandedness is maintained during library preparation
Detect which strand of the genome encodes the gene transcript and differentiate between two distinct alleles encoded on opposite strands of the genome.
Download
Visual feedback at each step in the workflow
For highest success rate, tracking dyes change color at each step only when proper mixing is achieved. Lack of a color change indicates the need to pause and complete mixing prior to continuing. Visual feedback is useful to monitor the performance of robotic systems.
Video: Collibri NGS Library Prep kits for Illumina systems
Video Player is loading.
Current Time 0:00
Duration 1:10
Loaded: 0.00%
Stream Type LIVE
Remaining Time 1:10
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Tips for your RNA-seq library preparation
    The scientists who developed the Collibri Stranded RNA Library Prep kits share their tips for cDNA library generation which retains the full transcriptome.
    Recommended controls
    ERCC RNA Spike-In Mixes are recommended to be added to the input RNA before library preparation. The mixes (not included in the kit) provide a set of external RNA controls that enable performance assessment for dynamic range, lower limit of detection, and fold-change response for gene expression experiments. For detailed information, refer to the ERCC RNA Spike-In Control Mixes User Guide.
    For positive RNA control, we recommend using Universal Human Reference RNA (Cat. No. QS0639) or Human Brain Total RNA (Cat. No. AM7962).
    Tip!
    Do not use the ERCC RNA Spike-In Control Mixes with highly degraded samples such as FFPE RNA. High quality ERCC RNA can outcompete lower quality RNA population thus causing ERCC to be overrepresented in the final library.
    Whole-transcriptome library preparation workflow
    Retain coding and noncoding RNA diversity
    We offer a range of Invitrogen genomic RNA extraction kits for sensitive, scalable purification from an expansive set of starting materials to help maximize process efficiency and downstream performance. This includes a broad range of kits for purifying RNA from a variety of samples including tissue, cells, blood, serum, plants, forensic samples, and more.
    Automate RNA extraction using magnetic bead-based technology and KingFisher instruments to bind, wash, and elute RNA to reduce your hands-on time while maintaining high yields and excellent reproducibility.
    Ribosomal depletion for bacteria or plants
    The RiboMinus Isolation kits enable efficient and selective removal of ribosomal RNA (rRNA) from intact total RNA in less than two hours and are compatible with Collibri Library Prep kits.
    Recommended accessories
    Quantification options to scale throughput
    The qPCR-based Collibri Library Quantification kit scales well for larger sample batches and is the ideal method for precious samples or clinical samples. Quantification accuracy is equivalent to the KAPA Library Quantification kit with the additional benefit of visual feedback during the quantification process.
    The Qubit dsDNA HS Assay is a fluorometric assay that uses dsDNA-binding dyes in order to accurately determine NGS library concentration, and benefits from a simple workflow of just a few minutes per sample. 
    Regardless of the assay that is chosen, good laboratory technique should be used in order to ensure accurate measurement of library concentrations and high-quality Illumina sequencing data.
    Recommended accessories
    With Applied Biosystems QuantStudio real-time PCR systems, you get true value with excellent performance, reliability, and world-class support. Our family of instruments enables you to obtain the results you need, connect and collaborate with colleagues, and achieve your research goals.
    Retain coding and noncoding RNA diversity
    We offer a range of Invitrogen genomic RNA extraction kits for sensitive, scalable purification from an expansive set of starting materials to help maximize process efficiency and downstream performance. This includes a broad range of kits for purifying RNA from a variety of samples including tissue, cells, blood, serum, plants, forensic samples, and more.
    Automate RNA extraction using magnetic bead-based technology and KingFisher instruments to bind, wash, and elute RNA to reduce your hands-on time while maintaining high yields and excellent reproducibility.
    Ribosomal depletion for bacteria or plants
    The RiboMinus Isolation kits enable efficient and selective removal of ribosomal RNA (rRNA) from intact total RNA in less than two hours and are compatible with Collibri Library Prep kits.
    Recommended accessories
    Quantification options to scale throughput
    The qPCR-based Collibri Library Quantification kit scales well for larger sample batches and is the ideal method for precious samples or clinical samples. Quantification accuracy is equivalent to the KAPA Library Quantification kit with the additional benefit of visual feedback during the quantification process.
    The Qubit dsDNA HS Assay is a fluorometric assay that uses dsDNA-binding dyes in order to accurately determine NGS library concentration, and benefits from a simple workflow of just a few minutes per sample. 
    Regardless of the assay that is chosen, good laboratory technique should be used in order to ensure accurate measurement of library concentrations and high-quality Illumina sequencing data.
    Recommended accessories
    With Applied Biosystems QuantStudio real-time PCR systems, you get true value with excellent performance, reliability, and world-class support. Our family of instruments enables you to obtain the results you need, connect and collaborate with colleagues, and achieve your research goals.