Shi Asymmetric Epoxidation – an electrophilic addition reaction
There had been many previous attempts to create an efficient non-metal catalyst for asymmetric epoxidations. In 1996, Yian Shi of Colorado State University developed a fructose-derived ketone catalyst that demonstrated excellent enantioselectivity. Since this discovery, the use of Shi’s catalyst has become known as the Shi asymmetric epoxidation.
The Shi epoxidation involves treating alkenes with oxone (potassium peroxymonosulfate) in the presence of the Shi catalyst. The reaction is believed to proceed via a dioxirane intermediate generated from the ketone catalyst by the oxone. One potential side reaction that can occur is the Baeyer-Villiger oxidation, where rearrangement of the peroxy group results in the formation of an ester. However, this can be mitigated by maintaining the pH at an optimum value of approximately 10.5.
The Shi epoxidation has been used in the key step of several total synthesis campaigns, including the synthesis of glabrescol, a chiral C2-symmetric pentacyclic oxasqualenoid, by Elias J. Corey and colleagues.
Review available Thermo Scientific products for the Shi asymmetric epoxidation reaction:
Catalog # Name Size Price (USD) Qty
L18989.03 D-Epoxone, 98% Each
50.65
Special offer
Online exclusive
Ends: 30-Jun-2025
59.60
Save 8.95 (15%)
L18989.06 D-Epoxone, 98% Each
191.65
Online Exclusive
213.00
Save 21.35 (10%)
Vote for your favorite named reaction
Learn more about the most popular named reactions based on feedback from our customers.  Do you have a favorite reaction?  Let us know! 
Vote now