Watch this short video for an overview of warning signs to diagnostic investigations, so you can help improve earlier diagnosis:
Dr Joseph Mikhael6, MD and Chief Medical Officer, International Myeloma Foundation recommended several blood tests to rule out Multiple Myeloma, in his recent publication . These include, but are not limited to:
Complete blood count to check for anemia and raised Erythrocyte Sedimentation Rate (ESR)
Chemistry to check for raised calcium, raised creatinine and low albumin
Serum free light chain (sFLC) tests, serum protein electrophoresis (SPE) and immunofixation (sIFE) to check for a monoclonal protein
If an MGUS diagnosis is confirmed by hematologists, primary care providers may be asked to monitor these patients.
While MGUS is not a malignant condition, it has significant clinical implications. Patients with MGUS have an increased risk of progression to hematological malignancies8, such as Multiple Myeloma, as well as being more susceptible to infections9, renal impairment10, osteoporosis and fractures11, neurological disorders12 and thrombosis13.
Regular monitoring of these patients helps with early detection and treatment of more serious conditions that may develop.