Choose the NanoDrop instrument that fits your needs
We offer a choice of instruments to fit the needs of most laboratories. Whether it’s our flagship Thermo Scientific NanoDrop Ultra Microvolume UV-Vis Spectrophotometers for quantification, purity, and sample intelligence; our high-throughput NanoDrop Eight Spectrophotometer to process plated samples, eight at a time; or our basic NanoDrop Lite Plus Spectrophotometer for essential quantitation and purity ratios, we have an instrument to suit your lab’s applications and budget.
Current promotions
Save on the latest NanoDrop and other Thermo Scientific UV-Vis instruments—especially if you are upgrading from an older model.
Which NanoDrop instrument should you choose?
NanoDrop Ultra/UltraC/
UV-Vis
NanoDrop Eight
UV-Vis
NanoDrop Lite Plus 
UV
Product summary
Full-spectrum microvolume UV-Vis measurements, use a standard cuvette, and/or measure highly concentrated samples more accurately with the Acclaro Pro software.
High-throughput, full-spectrum microvolume UV-Vis measurements
Basic microvolume UV measurements
Sampling options
Pedestal (All Models)
Cuvette (NanoDrop UltraC)
Pedestal
Pedestal
Measures 1–2 µL sample
Dynamic range
(ng/µL dsDNA)
1.0–27,500*
ng/µL for dsDNA
2.0–10,000
ng/µL for dsDNA
2–1,500
ng/µL for dsDNA
Full-spectra data
Pre-programmed methods for nucleic acids and proteins
Measures nucleic acid A260
Measures nucleic acid A260/A280 ratio
Measures nucleic acid A260/A230 ratio
Measures purified protein A280
Measures protein and peptides at A205
Measures protein A260/A280 ratio
Pre-programmed methods for colorimetric assays (BCA, Bradford, Lowry, Pierce 660)
Custom methods
Built-in cuvette option
 (NanoDrop UltraC)
Standalone local control
Requires computer
High-resolution touchscreen interface
Integrated qPCR Recipe Calculator    
Measures fluorescent-labeled nucleic acid and protein samples using absorbance
Compatible with the NanoDrop Ultra dsDNA and RNA Fluoresence Assays
Capable of taking fluorescence measurements
Autoblank and Automeasure
Acclaro contaminant identification
Acclaro Pro Compatibility    
21 CFR Part 11 regulatory compliance
Optional SciVault 2 Software
Optional SciVault Software
Password-protected access
Optional SciVault 2 Software
Optional SciVault Software
Embedded technical support **
Enhanced connectivity (Wi-Fi and Bluetooth™)
Cloud Compatibility
 (Microsoft OneDrive, Google Drive, Thermo Fisher Connect)
LIMS API Support    
*Lowest detection limit is 0.2 ng/µL with 10 mm cuvette.
**Instrument is delivered with English software and help. Language packs can be downloaded from our website
NanoDrop Ultra Microvolume UV-Vis Spectrophotometers
The Thermo Scientific NanoDrop Ultra Spectrophotometers are compact, stand-alone UV-visible spectrophotometers for measurement of highly concentrated samples without the need for dilution across a wide variety of analytes. A glove-compatible touchscreen offers easy access to preloaded software applications and enables purity and concentration assessments in seconds. Built-in Thermo Scientific Acclaro Sample Intelligence technology helps identify and correct for the contaminants present.
The NanoDrop Ultra introduces a suite of innovative features designed to enhance your laboratory workflow. Key highlights include an integrated qPCR calculator that precisely determines the sample volume required for downstream applications, and the optional Acclaro Pro software, which incorporates an advanced algorithm for highly accurate quantification of concentrated samples. Additionally, a diagnostic scheduler alerts you when a diagnostic test is due, ensuring consistent performance. These, along with numerous other cutting-edge features, make the NanoDrop Ultra an indispensable tool for modern laboratories.
NanoDrop Eight Microvolume UV-Vis Spectrophotometer
The compact Thermo Scientific NanoDrop Eight Spectrophotometer measures eight samples at a time in less than 20 seconds. It is an ideal choice for pharma/biotech and academic researchers needing a higher throughput capacity for measuring nucleic acid and protein samples. NanoDrop Eight software identifies sample contaminants and reports true sample concentration with the built-in Thermo Scientific Acclaro Sample Intelligence technology.
NanoDrop Lite Plus Spectrophotometer
The affordable Thermo Scientific NanoDrop Lite Plus Microvolume Spectrophotometer enables any lab to easily assess purified DNA, RNA, and protein concentrations and purity ratios. While this entry-level unit offers fewer features than our premium models, it delivers the same fast, accurate, reliable measurements synonymous with the NanoDrop platform. The easy-to-use, affordable package is a great option for teachers and researchers.
NanoDrop spectrophotometers and fluorometers vs Qubit fluorometer
In addition to UV-Vis absorbance, another technology often used to quantify nucleic acids or proteins is fluorometry, in which samples are stained with a fluorescent dye highly specific to the analyte of interest. Invitrogen Qubit Fluorometers use this method to quantify DNA, RNA, and protein.
Which is better? It depends on your application and the needs of your laboratory. If you need sample qualification or purity ratios, you’ll want a NanoDrop spectrophotometer. If you have very dilute or difficult-to-process samples, you'll want a Qubit fluorometer. In some circumstances, you may want both.
The table shows key differences between the two technologies. For a more comprehensive comparison, see our RNA/DNA quantification page.
Instrument NanoDrop Ultra/UltraC spectrophotometers Qubit 4 fluorometer
Technology UV-Vis absorbance Fluorescence
Key advantages − Fast and simple. Allows for direct quantification of samples without dilution or sample preparation.
Identifies and corrects for contaminants, including RNA in DNA samples and vice versa. Allows for direct quantification of samples without dilution or sample preparation.
− Identifies and corrects for contaminants, including RNA in DNA samples and vice versa.
− Sensitive. Accurate for very dilute or difficult-to-process samples. Most sensitive of all three options.
− Specific. Can distinguish between dsDNA and ssDNA or microRNA and total RNA. Assays kits contain reagents specific to many common analytes.
Time to results
Seconds
Minutes
dsDNA quantitation range 0.2–27,500 ng/µL* 0.005–4,000 ng/µL †
Sample qualification (purity) − Calculates standard purity ratios.
− Acclaro Sample Intelligence technology identifies and corrects for contaminants such as protein or guanidine.
− Assays are relatively tolerant of common contaminants but do not identify or quantify them.
  *dsDNA absorbance quantitation range for NanoDrop Ultra/UltraC instrument is 1.0 ng/µL with pedestal and 0.2 ng/µL with cuvette.
  **NanoDrop Ultra dsDNA HS Fluorescence Assay kits can quantify samples as dilute as 0.1 ng/µL, while the NanoDrop Ultra dsDNA BR Fluorescence Assay kit can measure samples as concentrated as 1,000 ng/µL, depending on sample to reaction volume.
  Qubit HS and 1X dsDNA Assay kits can quantify samples as dilute as 0.005 ng/µL, while the Qubit 1X dsDNA BR Assay kit can measure samples as concentrated as 4,000 ng/µL, depending on sample volume.
When would you want to use both absorbance and fluorescence measurements?
If you need both highly sensitive nucleic acid or protein quantitation and assessments of sample purity, a NanoDrop Ultra Spectrophotometer and Fluorometer would be a great fit for your lab.
For example, if your lab workflow includes qPCR assays, you will need accurate template quantitation and contaminant identification to comply with MIQE guidelines, required by many scientific journals. As shown in the video, insufficient template can lead to inaccurate results. Using a NanoDrop Ultra Spectrophotometer and Fluorometer can give you a more complete picture of input RNA quantity, purity, and integrity.
For customers working in higher throughput environments, the combination of the NanoDrop Eight and Qubit Flex offer the same level of detail needed but with the added ability to process 8 samples at a time.
For Research Use Only. Not for use in diagnostic procedures.