Search Thermo Fisher Scientific
Make sure you’re operating your Thermo Scientific NanoDrop Microvolume Spectrophotometers and Fluorometers with full functionality. Based on input from customers like you, we are constantly updating and evolving NanoDrop software to deliver greater capabilities and efficiencies in the lab.
You can download and install the latest operating software by following the steps in this section. Minimum computer and operating system requirements needed to run each NanoDrop instrument are described in the installation section.
Tip: If you are using a NanoDrop Ultra or NanoDrop Eight instrument in a regulated environment, you can also purchase optional compliance software that helps you meet FDA 21 CFR Part 11 requirements for electronic recordkeeping. Visit the NanoDrop Ultra or NanoDrop Eight catalog page for more information or simply order below.
Registration is a one-time process that allows you to receive alerts when new software is available for your instrument, and helps verify your warranty status.
When you register, make sure to check the box allowing marketing emails from Thermo Fisher Scientific, as we cannot alert you to new updates without your permission.
The table lists the latest operating software versions available for your instrument. To check your current version, select System status (on your NanoDrop Ultra/UltraC/Ultra FL/UltraC FL, NanoDrop One/OneC or NanoDrop Eight instrument) or select Help > About from the menu in the software on your PC (for all other instruments).
Latest operating software for NanoDrop instruments
Instrument |
Software module |
Version |
Release date |
Download |
---|---|---|---|---|
NanoDrop Ultra/UltraC/Ultra FL/UltraC FL |
Local Control Software | 1.0 |
February 2025 | |
PC Control Software | 1.0 | February 2025 | ||
NanoDrop Eight |
Operating Software |
1.6 |
December 2023 | |
NanoDrop Lite Plus |
Operating Software |
1.2.1 |
November 2023 |
|
NanoDrop One/OneC | Local Control Software | 2.12.0 | October 2024 | |
PC Control Software | 2.12.0 |
October 2024 | ||
NanoDrop 3300 |
Operating Software |
2.8.1 |
August 2020 |
|
NanoDrop 2000/2000c |
Operating Software |
1.6 |
September 2014 |
|
NanoDrop 8000 |
Operating Software |
2.3.3 |
April 2020 |
|
NanoDrop 1000 |
Operating Software |
3.8.1 |
June 2011 |
To download the latest version:
Once the file(s) are downloaded to your computer, come back to this page and refer to the appropriate section for installation instructions.
Because the NanoDrop Ultra spectrophotometers and fluorometers can be controlled from either its local touchscreen or from a connected PC, you will need to install software update files for both.
Important: To ensure data transfer and viewing, the software versions for local instrument and PC control must match. For example, if you are updating your local NanoDrop Ultra Operating and help software to version 1.0 (February 2025), make sure to update your PC control software to version 1.0 (February 2025) as well.
Install local control software on your NanoDrop Ultra/UltraC/Ultra FL/UltraC FL instrument via a USB drive or through an online update.
NanoDrop Ultra HELP system file (March 2025)
Through USB:
Through Online Update:
(Optional) Install a custom method on your NanoDrop Ultra/UltraC/Ultra FL/UltraC FL instrument via a USB drive.
For Research Use Only. Not for use in diagnostic procedures.