Search Thermo Fisher Scientific
All Thermo Scientific NanoDrop microvolume spectrophotometers include our innovative technology which allows you to quantify DNA, RNA, and protein from just 1–2 µL of sample. With no need for cuvettes or capillaries, results are delivered in seconds, streamlining your workflow and enhancing your research capabilities.
The NanoDrop Ultra offers exceptional accuracy for advanced applications, while the NanoDrop Eight maximizes throughput by measuring eight samples simultaneously. The NanoDrop Lite Plus provides a compact, user-friendly solution for routine measurements. Trusted by researchers worldwide and cited in over 55,000 scientific publications, NanoDrop spectrophotometers are indispensable tools in any lab, ensuring reliable and rapid results every time.
Gain the most comprehensive information about the concentration and quality of your DNA or RNA samples with the innovative NanoDrop Ultra FL and NanoDrop UltraC FL Spectrophotometers, which uniquely combine microvolume UV-Vis absorbance and fluorescence measurements into a single, versatile platform.
For Research Use Only. Not for use in diagnostic procedures.