The detection and quantitation of proteins are vital to many biological studies, due to the ubiquitous and fundamental roles of proteins in biological processes as well as the commercial importance of proteins in the biotech and pharmaceutical industries.
There are currently several absorbance- and fluorescence-based protein assays in common use, each with its own shortcomings, such as protein-to-protein variability, contaminant interference, time requirements, accuracy, sensitivity, and the need for caustic or hazardous reagents. We compared four common protein assays for protein-to-protein variation, accuracy, precision, and sensitivity. The Quant-iT Protein Assay for the Qubit Fluorometer compared favorably by providing low protein variability, rapid quantitation, accuracy, precision, and high sensitivity.
NOTE: Since this article was published, we have launched the next generation instrument,  the Qubit 2.0 Fluorometer which quantitates DNA, RNA, and protein with unprecedented accuracy, sensitivity, and simplicity.
The Qubit Quantitation Platform: Fast and Easy to Use
The Qubit Quantitation Platform is the combination of a user-friendly fluorometer with highly sensitive fluorescence-based quantitation assays. The Qubit Fluorometer is a small, economical instrument designed to work seamlessly with Quant-iT Assay Kits for routine protein, DNA, and RNA quantitation (Figure 1). All settings and calculations are performed for you. The system is simple, fast, and easy to use, yet consistently produces accurate results, so you can be confident moving forward with subsequent applications. Each Quant-iT Assay Kit is highly specific for a single analyte, and all are more sensitive than absorbance-based measurements. Only small sample volumes of 1–20 μL are required, and all assay reagents are stored at room temperature, eliminating the need to thaw reagents.
Workflow for Quant-iT™ assays with Qubit® Fluorometer
Figure 1. Workflow for the Quant-iT assays using the Qubit Fluorometer.
Quant-iT Protein Assay: Accurate, Precise, and Sensitive
We compared 3 assays to the Quant-iT Protein Assay for the Qubit Fluorometer (Figure 2): the Bio-Rad® Quick Start™ Bradford Protein Assay, the Pierce BCA Protein Assay, and the Pierce Modified Lowry Protein Assay. The Bradford method (using Coomassie Brilliant Blue) [1] exhibited very high protein-to-protein variability (Figure 2B). Spectrophotometric assays such as those using BCA (bicinchoninic acid) [2] require carefully timed steps, are not compatible with reducing agents, and often yield high estimates of protein, as observed for lysozyme in our study (Figure 2C). The Lowry method [3] (Figure 2D) employs a lengthy, multistep procedure and is incompatible with detergents, carbohydrates, and reducing agents. The Quant-iT™ Protein Assay showed low protein-to-protein variation, good accuracy and precision, and sensitivity down to 0.025 mg/mL in the stock sample (Figure 2A).
The Quant-iT Protein Assay is insensitive to many common contaminants, including reducing agents, nucleic acids, and free amino acids. However, detergents, such as SDS (final concentration >0.01%), Tween® 20, and Triton® X-100, are not recommended. The assay has an optimal range of 1.25 to 25 µg/mL (0.25–5 µg) in the assay tube (initial stock concentrations, 12.5–5 mg/mL) and is provided in a simple kit format that allows easy and rapid use. Learn more about the Qubit Quantitation Platform.
Quant-iT Protein Assay
Figure 2. The Quant-iT™ Protein Assay with the Qubit Fluorometer produces less protein-to-protein variation and higher accuracy, precision, and sensitivity than 3 other common protein assays. (A–D) The same lot of each protein was used in all assays, and assays were carried out in triplicate following the manufacturers’ protocols. Data are graphed to show protein-to-protein variation throughout the protein concentration range tested. The inset in (A) is a magnification of the low end of the protein range, to show the sensitivity of the Quant-iT Protein Assay used with the Qubit Fluorometer.
Ordering Information
Catalog # Name Size Price (USD) Qty
Q33211 Qubit™ Protein Assay Kit, 100 Assays Each
149.65
Online Exclusive
158.00
Save 8.35 (5%)
Q33212 Qubit™ Protein Assay Kit, 500 Assays Each
447.65
Online Exclusive
476.00
Save 28.35 (6%)
Q10210 Qubit™ RNA BR Assay Kit, 100 assays Each
139.65
Online Exclusive
145.00
Save 5.35 (4%)
Q10211 Qubit™ RNA BR Assay Kit, 500 assays Each
408.65
Online Exclusive
431.00
Save 22.35 (5%)
Q32852 Qubit™ RNA HS Assay Kit, 100 assays Each
139.65
Online Exclusive
145.00
Save 5.35 (4%)
Q32855 Qubit™ RNA HS Assay Kit, 500 assays Each
406.65
Online Exclusive
430.00
Save 23.35 (5%)
Q10212 Qubit™ ssDNA Assay Kit Each
146.00
Q32850 Qubit™ dsDNA BR Assay Kit, 100 assays Each
147.00
Q32853 Qubit™ dsDNA BR Assay Kit, 500 assays Each
435.00
Q32851 Qubit™ dsDNA HS Assay Kit, 100 assays Each
147.00
Q32854 Qubit™ dsDNA HS Assay Kit, 500 assays Each
437.00
Q32856 Qubit™ Assay Tubes Each
128.00
Quick Product View
See a complete listing of the products discussed in this article.
View products
Learn More
Find more information on the Qubit fluorometer and fluorescence-based Quant-iT assays.
Download This Article
Get a copy of this article as it appears in the print version of BioProbes 63.
For Research Use Only. Not for use in diagnostic procedures.