Plastic beakers
Rely on plastic beakers for a safer alternative to glass, especially in larger sizes.
They won’t break, chip or shatter during use and cleaning. Lightweight and easy to handle. Chemical resistance, heat tolerance (not for use on hot plates), and spout design make Thermo Scientific Nalgene Griffin low-form beakers a perfect long-term solution for your graduated beaker choice. Meeting the ISO/DIN 7056 International Standards for Laboratory Plastic Beakers, our polypropylene and polymethylpentene beakers enable precise measurements. Nalgene beakers with handles offer reliable support and dispensing control. Nalgene beakers are made only from high quality laboratory grade plastic materials for dependably lower levels of potential extractables and leachables (E&L), so you can rely on them for everyday lab use without interfering with your work. 
Looking for ways to make your lab more sustainable?  Nalgene polypropylene copolymer beakers are reusable, durable and  recyclable at end-of-life in many communities; discover more about the sustainability of using reusable plastic labware and Thermo Fisher Scientific's green manufacturing practices through our participation in My Green Lab's ACT Label program.
Which beakers are best for your applications?
Nalgene PPCO Griffin Low-Form Plastic BeakersNalgene PMP Griffin Low-Form Plastic Beakers 
Nalgene PFA Griffin
Low-Form Plastic Beakers
Nalgene HDPE Graduated Plastic Beakers with HandlesNalgene PMP Graduated Plastic Beakers with HandlesNalgene PFA Graduated Plastic Beakers with Handles
product-beakerproduct-beaker-pmpproduct-beaker-lfproduct-beaker-hdpeproduct-beaker-hdi-pmpproduct-beaker-pfa
Milky white, graduated, good chemical resistanceTransparency of glass, graduated, excellent chemical resistance, heat toleranceMilky white, heavy wall, ultimate chemical resistance, heat toleranceMilky white, extra-heavy wall, sturdy rectangular handle, reliable support, dispensing controlTransparent, graduated, sturdy rectangular handle, easy, safe handling and pouringMilky white, heavy wall, ultimate chemical resistance, heat tolerance
Meets ISO/DIN 7056**, 21 CFR Pt 177.1520Meets ISO/DIN 7056"", 21 CFR Pt 177.1520Meets 21 CFR Pt 177.1520Meets 21 CFR Pt 177.1520
AutoclavableAutoclavableAutoclavableNot autoclavableAutoclavableAutoclavable
Max use 121oC*Continuous use 150oC intermittent use up to 170oC*Max Use 250oC*Max use 120oC*
Continuous use 150oC intermittent use up to 170oC*Max Use 250oC*
Graduations:
silk screened
Graduations:
silk screened
No graduationsGraduations:
molded-in;
oz and mL
Graduations:
molded-in;
oz and mL
Graduations:
molded-in;
oz and mL
Volume range:
30mL–4L
Volume range:
50mL–4L
Volume range:
50mL–1L
Volume range:
1L, 3L
Volume range:
1L, 3L
Volume range:
1L, 3L
PPCOPMPPFAHDPEPMPPFA
Translucent whiteClearTranslucent whiteTranslucent whiteClearTranslucent white
No handleNo handleNo handleHandleHandleHandle
NestingNestingNestingNot nestingNot nesting Not nesting
BPA freeBPA freeBPA freeBPA freeBPA freeBPA free
Phthalate-freePhthalate-freePhthalate-freePhthalate-freePhthalate-freePhthalate-free
ACT Label ›     
Order NowOrder NowOrder NowOrder NowOrder NowOrder Now
*Nalgene beakers are not for use on hot plates.
**International Standards for Laboratory Plasticware
General chemical compatibility+
 Nalgene
PPCO Griffin Low-Form
Plastic Beakers
Nalgene
PMP Griffin
Low-Form
Plastic Beakers
Nalgene
Griffin
Low-Form PFA
Plastic Beakers
Nalgene Graduated HDPE
Plastic Beakers
with Handles
Nalgene PMP Graduated Plastic Beakers with HandlesNalgene PFA Graduated Plastic Beakers with Handles
Acids, dilute or weak
Acids^ strong, concentrated
Alcohols, aliphatic
Aldehydes
Bases/alkali
Esters
Hydrocarbons, aromatic
Hydrocarbons, halogenated 
Ketones, aromatic
Oxidizing agents, strong
+ Ratings are for short term intermittent exposure. Wash thoroughly between uses.
^ Except oxidizing acids; for oxidizing acids, see “Oxidizing agents, strong.”
Thermo Scientific Nalgene Economy Beakers are a simplified version of the original Nalgene premium beakers for those with more basic product requirements. The Economy Beakers are made from high quality laboratory grade polypropylene copolymer for reliably low extractables and shatter-proof performance at temperatures above -40°C, just like Nalgene premium beakers. The molded in-nominal graduations found on the Economy Beakers differ from the premium beaker’s high visibility printed graduations and, thus, reduces cost
Learn more
Start every day with dependable labware | Thermo Scientific Nalgene labware
Your work depends on quality and consistency, and we believe every lab should have the best tools. Watch Video
Video Player is loading.
Current Time 0:00
Duration 1:46
Loaded: 0%
Stream Type LIVE
Remaining Time 1:46
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    ACT
    : Accountability. Consistency. Transparency.
    When looking for sustainable products, we believe it should be easy to make informed purchasing decisions. We're working to provide more information about our products' environmental impact by participating in the ACT Label program with a selection of Thermo Scientific Nalgene and Matrix products. It's like an eco-nutrition label for sustainability.
    Learn more
    Resources
    Certificates of Compliance
    Receive a Certificate of Compliance and Certificate(s) of Irradiation (if applicable) on purchased Thermo Scientific Nalgene and Nunc products.
    Start your request 
    Support
    Need technical support? Contact our expert team for technical and application support of Laboratory Products at technicalsupport@thermofisher.com.
    California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.