Organism TypeMouseEmbryonic mouse fibroblast; Properties: contact inhibition; transformation; transfection; cell biology; ability to differentiate into adipocytes. Susceptible to: murine leukemia virus, murine sarcoma virus, vesicular stomatitis, vaccinia, herpes simplex, N-tropic oncornaviruses C
TissuesFibroblast
PhenotypeAdherent
Primaryno
ApplicationRNAi,Transfection: Gene Expression,Transfection: Gene Silencing,Transient Transfection
Recommended Transfection Reagents
  • Plasmid DNA transfection and co-transfection: Lipofectamine® 3000 (51-79% transfection efficiency and 4 fold protein expression improvement over Lipofectamine® 2000)
  • RNAi transfection (siRNA/miRNA) and gene silencing: Lipofectamine® RNAiMAX
Products
Catalog # Name Size Price (USD) Qty
11668019 Lipofectamine™ 2000 Transfection Reagent, 1.5 mL Each
842.65
Online Exclusive
924.00
Save 81.35 (9%)
13778075 Lipofectamine™ RNAiMAX Transfection Reagent, 0.75 mL Each
634.65
Online Exclusive
676.00
Save 41.35 (6%)
13778150 Lipofectamine™ RNAiMAX Transfection Reagent, 1.5 mL Each
1,040.65
Online Exclusive
1,108.00
Save 67.35 (6%)
15338100 Lipofectamine™ LTX Reagent with PLUS™ Reagent, 1 mL Each
854.65
Online Exclusive
940.00
Save 85.35 (9%)
18292011 Lipofectin™ Transfection Reagent, 1 mL Each
710.00
18324012 Lipofectamine™ Transfection Reagent, 1 mL Each
682.00
Every step of the way, a wide range of cell health products
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.