Generate high-quality Sanger sequencing data with our proven workflow
Our simple and fast Sanger sequencing workflow can be completed in less than one workday, from sample to answer. Thermo Fisher Scientific offers Applied Biosystems products to support many steps of our recommended workflow, from PCR amplification to data analysis. Learn more about each step of our recommended Sanger sequencing workflow below.
Video: How to Perform Sanger Sequencing
Video Player is loading.
Current Time 0:00
Duration 2:56
Loaded: 0%
Stream Type LIVE
Remaining Time 2:56
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Workflow
    COL13523-consumables-workflow
     Primer Designer Tool for PCR & Sanger Sequencing
    Use our online Invitrogen™ Primer Designer™ Tool to search for the right PCR/Sanger sequencing primer pair from a database of ~650,000 predesigned primer pairs for resequencing the human exome and human mitochondrial genome. Choose from different amplicon lengths to accommodate various research applications and biological sample types.
    • Our primers are free of known SNPs and primer-dimers, highly target-specific and used under universal PCR conditions 
    • Full primer coverage for Ion Torrent™ AmpliSeq™ Exome Panel and Ion Torrent™ AmpliSeq™ Cancer Hotspot Panel v.2 Sanger confirmation workflow
    • Flexible primer configuration to meet your research needs: primers can be ordered unmodified, M13-tailed, HPLC-purified or desalted
    • All the primers have been checked by mass spectrometry and have passed stringent bioinformatics metrics; lab bench validation tests have shown >95% success rate
    Visit tool ›
    Platinum II Taq Hot-Start DNA Polymerase
    Invitrogen Platinum II Taq Hot-Start DNA Polymerase helps you get to sequencing reactions faster. A unique combination of innovative buffer, high-performance Taq DNA polymerase, and superior Platinum hot-start technology helps enable exceptional PCR results, even in the toughest applications.
    • Universal primer annealing at 60°C enables co-cycling of all assays, reducing tedious optimization steps
    • An engineered Taq polymerase enables 4X faster DNA synthesis, inhibitor resistance, and robust amplifi cation
    • Platinum hot-start technology offers superior specificity, sensitivity, and yields, and allows for room temperature reaction setup
    • 2X master mix format helps reduce pipetting errors with fewer pipetting steps
    More formats and information are available at thermofisher.com/platinumiitaq.
    Catalog # Name Size Price (USD) Qty
    14000012 Platinum™ II Hot-Start PCR Master Mix (2X), 50 reactions Each
    106.65
    Online Exclusive
    111.00
    Save 4.35 (4%)
    14000013 Platinum™ II Hot-Start PCR Master Mix (2X), 200 reactions Each
    389.65
    Online Exclusive
    412.00
    Save 22.35 (5%)
    14000014 Platinum™ II Hot-Start PCR Master Mix (2X), 1000 reactions Each
    1,544.65
    Online Exclusive
    1,650.00
    Save 105.35 (6%)
    ExoSAP-IT Express reagent: fastest PCR cleanup method 
    The Applied Biosystems ExoSAP-IT Express reagent offers rapid turnaround times and improvedefficiency of resource use while delivering the same superior cleanup as the original ExoSAP-IT reagent. Eliminate spin columns, magnetic beads, sedimentation, filtration, and gel purification. The novel ExoSAP-IT technology allows for a significant reduction in sample cleanup time with minimal steps, providing the simplest workflow.
    • 5 min protocol
    • One-tube, one-step PCR cleanup
    • Novel enzyme technology
    • 100% recovery of PCR products
    EM5034479-ExoSAP-IT pic
    Video: How to Clean up PCR Reactions with ExoSAP-IT Express Reagent
    Video Player is loading.
    Current Time 0:00
    Duration 0:55
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:55
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Catalog # Name Size Price (USD) Qty
      75001.200.UL ExoSAP-IT™ Express PCR Product Cleanup Reagent, 100 reactions Each
      165.65
      Online Exclusive
      172.00
      Save 6.35 (4%)
      75001.1.ML ExoSAP-IT™ Express PCR Product Cleanup Reagent, 500 reactions Each
      648.65
      Online Exclusive
      676.00
      Save 27.35 (4%)
      75001.4X.1.ML ExoSAP-IT™ Express PCR Product Cleanup Reagent, 2000 reactions Each
      2,313.65
      Online Exclusive
      2,435.00
      Save 121.35 (5%)
      75001.10.ML ExoSAP-IT™ Express PCR Product Cleanup Reagent, 5000 reactions Each
      5,248.65
      Online Exclusive
      5,490.00
      Save 241.35 (4%)
      We continue to develop superior Sanger sequencing kits that help provide solutions for all of your applications and templates.
      BigDye Direct Cycle Sequencing Kit
      The Applied Biosystems BigDye Direct Cycle Sequencing Kit simplifies the industry-standard Sanger sequencing workflow by combining post-PCR cleanup and cycle sequencing into a single step. The BigDye Direct Cycle Sequencing Kit requires PCR primers with M13 tails, which are available through the Primer Designer Tool.
      Video: Sequencing with BigDye Direct Cycle Sequencing Kit
      Video Player is loading.
      Current Time 0:00
      Duration 2:01
      Loaded: 0.00%
      Stream Type LIVE
      Remaining Time 2:01
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        bigdye_pic
        Catalog # Name Size Price (USD) Qty
        4458687 BigDye™ Direct Cycle Sequencing Kit, 100 reactions Each
        442.65
        Online Exclusive
        461.00
        Save 18.35 (4%)
        4458688 BigDye™ Direct Cycle Sequencing Kit, 1,000 reactions Each
        4,229.65
        Online Exclusive
        4,410.00
        Save 180.35 (4%)
        4458689 BigDye™ Direct Cycle Sequencing Kit, 24 reactions Each
        110.65
        Online Exclusive
        115.00
        Save 4.35 (4%)
        BigDye Terminator v3.1 Cycle Sequencing Kit
        Created by experts and refined by global teams, the BigDye Terminator v3.1 cycle sequencing kit offers exceptional versatility and high performance. This is a comprehensive, exhaustive solution, built for de novo sequencing, resequencing, and finishing with PCR product, plasmid, fosmid, and BAC templates.
        Improved quality of results and success rates, particularly with challenging templates
        • Suitable for a wide range of sequencing applications
        • Optimized for long read lengths
        • Sophisticated dye mobility characteristics
        • Refined performance reading through GT-rich regions
        • Optimal signal balance
        • Rapid completion of sequencing projects
        Learn more about BigDye Terminator v3.1 Cycle Sequencing Kit ›
        BigDye Terminator v1.1 Cycle Sequencing Kit
        The Applied Biosystems BigDye Terminator v1.1 Cycle Sequencing Kit is designed for specialty applications that require optimal basescalling adajcent to the primer, and for sequencing short PCR product templates with rapid electrophoresis run modules.
        Learn more about BigDye Terminator v1.1 Cycle Sequencing Kit ›
        BigDye XTerminator Purification Kit
        The Applied Biosystems BigDye XTerminator Purification Kit provides a fast, simple purification method for DNA sequencing reactions to remove unincorporated BigDye terminators and salts. No more dye blobs! Cleanup is complete in under 40 min and typically requires less than 10 min of labor.
        BDX-product-img
        Video: How to Purify Sequencing Reactions with the BigDye XTerminator Kit
        Video Player is loading.
        Current Time 0:00
        Duration 1:08
        Loaded: 0.00%
        Stream Type LIVE
        Remaining Time 1:08
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Catalog # Name Size Price (USD) Qty
          4376484 BigDye XTerminator™ Purification Kit, 2500 Preps Each
          4,899.65
          Online Exclusive
          5,130.00
          Save 230.35 (4%)
          4376485 BigDye XTerminator™ Purification Kit, 40,000 Preps Each
          29,251.65
          Online Exclusive
          30,630.00
          Save 1,378.35 (5%)
          4376486 BigDye XTerminator™ Purification Kit, 100 Preps Each
          362.65
          Online Exclusive
          380.00
          Save 17.35 (5%)
          4376487 BigDye XTerminator™ Purification Kit, 1000 Preps Each
          2,975.65
          Online Exclusive
          3,145.00
          Save 169.35 (5%)
          A range of platforms to meet your throughput needs
          Throughput
          Low
          Medium
          Medium
          High
          Learn moreLearn moreLearn moreLearn more
          On-instrument consumables are critical components of your sequencing run. Using the right Applied Biosystems products can help reduce your re-run rate and enables high-quality, reproducible data.
          SeqStudio-cartridge
          SeqStudio Cartridge
          The Applied Biosystems SeqStudio Cartridge is an easy-to-use reagent cartridge that includes capillaries, POP-1 universal polymer, buffer, and pump. The POP-1 universal polymer allows for flexibility to perform Sanger sequencing and fragment analysis on the SeqStudio Genetic Analyzer System with one cartridge. Just load your samples, click in the cartridge, and go.
          Browse other on-instrument consumables
          Video: How to set up a Sanger sequencing or fragment analysis run
          Video Player is loading.
          Current Time 0:00
          Duration 1:08
          Loaded: 0%
          Stream Type LIVE
          Remaining Time 1:08
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • en (Main), selected
            Minor Variant Finder Software
            The improved sensitivity achieved using the FREE Applied Biosystems Minor Variant Finder Software makes Sanger sequencing the ideal choice for oncologists and/or pathologists to call low frequency somatic variants (5% or below) where the number of relevant targets is often limited. The software requires no change to your current Sanger sequencing workflow.
            Benefits include:
            • Call minor variants at a detection level as low as 5%
            • Sequence a moderate number of targets at a low cost
            • Confirm next generation sequencing findings in alignment view and Venn diagram
            Video: How to detect minor variants using Sanger sequencing
            Video Player is loading.
            Current Time 0:00
            Duration 1:22
            Loaded: 0%
            Stream Type LIVE
            Remaining Time 1:22
            1x
              • Chapters
              • descriptions off, selected
              • captions off, selected
              • en (Main), selected
              Sanger Analysis Modules—free cloud-based tools
              Applied Biosystems Sanger Analysis Modules are FREE innovative cloud-based secondary data analysis tools that bring together multiple data sets in one convenient place. This online solution makes it easier to view, store, and analyze Sanger sequencing data.
              Key features include:
              • Next-generation sequencing (NGS) confirmation—one central location to confirm your NGS variants
              • Variant reporting in absolute genomic coordinates—eliminate the need to calculate from reference
              • Automated database search—automatically report genomic annotations for SNPs
              • .vcf output for downstream analysis—search multiple databases with the Ion Reporter annotation workflow
              More about Sanger sequencing and fragment analysis software ›
              Sanger Sequencing Kit
              The Applied Biosystems Sanger Sequencing Kit provides a convenient and affordable solution for preparing sequencing reactions. The kit provides all reagents needed for PCR cleanup, cycle sequencing, and sequencing product cleanup.     Learn more ›
              What makes a gold standard sequencing product?
              Gold standard represents quality and dependability. We commit to quality to help ensure results you can rely on.
              Learn more ›
              WB31610-362x250-SP_CXX
              Sanger sequencing chemistry guide
              A perfect handbook for scientists and researchers using Sanger sequencing. In this chemistry guide, you will learn the basic principles of Sanger sequencing from workflow to applications.
              • Sanger sequencing applications
              • DNA template preparation
              • Cycle sequencing
              • Purification of extension products
              • Capillary electrophoresis
              • Data analysis
              • Troubleshooting
              Build your knowledge
              Build your knowledge
              Get bite-sized answers to your everyday Sanger sequencing and fragment analysis questions
              Learn about the history of sequencing and how to pick the right platform for your research needs
              For Research Use Only. Not for use in diagnostic procedures.