Search Thermo Fisher Scientific
The Invitrogen Attune flow cytometer family offers a range of advanced features including high-speed sample acquisition, clog-resistant fluidics, flexible optical design, and exceptional sensitivity, setting a new standard in flow cytometry and empowering researchers to make their discoveries faster and with greater precision.
Explore how the capabilities of each model can enhance your lab's efficiency, accuracy, and overall research outcomes.
|
|
|
|
|
|
|
|
|
Try our instrument configurator tool to design an Attune NxT or CytPix system to fit your research needs.
Attune Flow Cytometers combine ultrasonic waves like those used in medical imaging with hydrodynamic forces to precisely position cells into a single, focused line in the central axis. Enabling cells to be tightly focused at the point of laser interrogation allows the system to collect more photons, helping to ensure data quality regardless of the sample-to-sheath ratio.
Acoustic focusing also lets your lab rapidly acquire high-quality data. Users can achieve sample throughput rates of 12.5 µL/min or 1,000 µL/min, up to 10 times faster than traditional hydrodynamic focusing systems and acquisition speeds of 35,000 events per second. This means processing all samples—including low-concentration and precious samples—more quickly and accurately with minimal loss in quality.
Rapid data acquisition
Compares the time required to acquire 1,000,000 events over three competitor instruments running at maximum sample rates.
Acoustic focusing minimizes variation regardless of the sample rate, so you don't have to make the tradeoff between throughput and sensitivity. This is demonstrated in the cell cycle analysis example below, where it's critical to precisely detect differences in fluorescence intensity between multiple cell populations.
Minimal data variation
Consistent results were achieved at high sample rates using Jurkat cells fixed and stained with propidium iodide, treated with RNase and analyzed at a concentration of 1 x 106 cells/mL. The coefficient of variation (CV) of cells in the G0/G1 and G2/M phases remain consistent, even at the highest sample rate of 1,000 μL/min.
To prevent clogging and allow for volumetric analysis, the Attune Flow Cytometers use a positive displacement syringe pump to control sample volume. The system can perform volumetric cell counts in a known volume (gated or total events) and can easily gate out dead cells to count live cells only in a live/dead analysis. Unlike traditional cytometers that operate at a maximum pressure of 15 PSI, Attune Flow Cytometers control pressure at 75 PSI, reducing the likelihood of cell-cell interactions and resulting in clog resistance.
Volumetric sample entry
Using positive displacement, the syringe pump works like a pipette. Tension is created with the plunger pushing down (A). The tension is released as the plunger rises with the sample (B). Clogging is less likely to occur under this high and controlled pressure environment.
Attune Flow Cytometers are configurable with up to 4 spatially separated lasers and 16 parameters. Spatial separation provides flexibility for multicolor panel design and streamlines compensation. The system offers superior speed with acquisition rates of up to 35,000 events per/second with high sensitivity to meet a range of research requirements.
High sensitivity distinguishes between dim signals and background, resulting in less variation and better signal separation. Fluorescent resolution coefficient variation is less than 3% for a single peak, and predicted MESF is ≤80 (FITC), ≤30 (PE), ≤70 (APC). Sensitivity in comparison to competitive systems is described below.
The compact size of the Attune Flow Cytometer also provides the flexibility of using it within a biosafety hood. This helps avoid contamination or infection when working with hazardous or unknown samples.
Attune Flow Cytometers feature a novel optical design that delivers first-class reliability and superior performance over time. The flat-top beam profile of the solid-state lasers minimizes the effects of changes in fluidics or optics, which in turn can lead to instability or alignment issues and instrument downtime.
Laser misalignment is a major concern with users of conventional flow cytometers. The flat-top lasers used in the Attune Flow Cytometers have an intensity profile that allows a wider window of alignment over Gaussian lasers used in traditional systems. The flat-top lasers also have a higher tolerance for misalignment that allows them to maintain high sensitivity and low CVs.
Emission profile of lasers
Gaussian laser used in traditional cytometers (left) with misalignment and flat-top laser used in the Attune Flow Cytometers (right) showing proper alignment if shifted.
Detection of rare events requires acquisition of high numbers of cells to attain a reliable measure of accuracy. Attune Flow Cytometers allow dilute samples to be processed quickly at sample input speeds of up to 1 mL/min, significantly faster than conventional cytometers that support maximum sample input rates of 60–100 µL/min. Acoustic focusing thus offers a unique combination of speed and quality, cutting the time to collect rare events significantly over long acquisition times.
Collecting more than 1 million live cells and detecting a rare population of dendritic cells
Plasmacytoid dendritic cells (pDCs) are identified using immunophenotype CD19–/B220high/CD317+. Four-color staining of mouse splenocytes included CD19-Pacific Blue, CD317-Alexa Fluor 488, CD45R/B220-PE direct conjugates, and SYTOX AADvanced Dead Cell Stain. A gate was made on live cells using SYTOX AADvanced Dead Cell Stain, followed by gating on CD19– cells. A two-parameter plot of CD45R/B220 vs. CD317 was used to identify pDCs. A collection rate of 500 μL/min was used to acquire 1.3 million total cells with a cell concentration of 7.5 x 107 cells/mL. Plasmacytoid dendritic cells were identified as dual B220+/CD317+ (upper right quadrant) and constitute 0.851% of live CD19– cells, which is 0.194% of total splenocytes.
Attune Flow Cytometers support both high speed of acquisition and high sensitivity to enable easy detection and phenotypic and functional characterization of rare cells. This step-by-step strategy for detecting rare events in our Flow Cytometry Learning Center will help you think through the best approach for pre-analytical, analytical, and data analysis phases of your research.
For Research Use Only. Not for use in diagnostic procedures.