Visualization of total protein and phosphoproteins in a 2D gel.
Visualization of total protein and phosphoproteins in a 2D gel. Proteins from a Jurkat T-cell lymphoma line cell lysate were separated by 2D gel electrophoresis and stained with Pro-Q Diamond phosphoprotein gel stain (P33300, P33301, P33302, blue) followed by SYPRO Ruby protein gel stain (S12000, S12001, S21900, red). After each dye staining, the gel was imaged on an FLA-3000 scanner (Fuji). The digital images were acquired using Z3 software (Compugen), and the resulting composite image was digitally pseudocolored and overlaid.
Visualization of total protein and phosphoproteins in a 2D gel. Proteins from a Jurkat T-cell lymphoma line cell lysate were separated by 2D gel electrophoresis and stained with Pro-Q Diamond phosphoprotein gel stain (P33300, P33301, P33302, blue) followed by SYPRO Ruby protein gel stain (S12000, S12001, S21900, red). After each dye staining, the gel was imaged on an FLA-3000 scanner (Fuji). The digital images were acquired using Z3 software (Compugen), and the resulting composite image was digitally pseudocolored and overlaid.
Related Products
Catalog # Name Size Price (USD) Qty
P33300 Pro-Q™ Diamond Phosphoprotein Gel Stain, 1 L Each
649.65
Online Exclusive
668.00
Save 18.35 (3%)
P33301 Pro-Q™ Diamond Phosphoprotein Gel Stain, 200 mL Each
200.65
Online Exclusive
206.00
Save 5.35 (3%)
P33302 Pro-Q™ Diamond Phosphoprotein Gel Stain, 5 L Each
2,738.65
Online Exclusive
2,815.00
Save 76.35 (3%)
S12000 SYPRO™ Ruby Protein Gel Stain, 1 L Each
445.65
Online Exclusive
465.00
Save 19.35 (4%)
S12001 SYPRO™ Ruby Protein Gel Stain, 200 mL Each
156.65
Online Exclusive
162.00
Save 5.35 (3%)
S21900 SYPRO™ Ruby Protein Gel Stain, 5 L Each
1,775.65
Online Exclusive
1,846.00
Save 70.35 (4%)
Related Images