Lab-Tek II RS Chamber Slide
Thermo Scientific Nunc Lab-Tek and Lab Tek II Chamber Slides allow you to seed, incubate, fix and stain on a single microscope slide with a removable chamber.
Which Chamber Slide is right for you?
Nunc Lab-Tek Chamber Slide System
Nunc Lab-Tek II Chamber Slide System
Nunc Lab-Tek II CC2 Chamber Slide System
Formats1, 2, 4, 8, 16 well1, 2, 4, 8 well1, 2, 4, 8 well
Chamber materialClear polystyrene with no handle to facilitate microscopic viewingClear polystyrene with top handle for aseptic manipulationsClear polystyrene with top handle for aseptic manipulations
Slide material
  • Permanox plastic, Nunclon Delta treated, or soda-lime glass, cell culture treated
  • Soda-lime glass, RS treated to aid cell attachment
  • Chemically modified CC2 surface to mimic poly-D-lysine
SealantMedical grade silicone, can be removed for coverslippingBiocompatible adhesiveBiocompatible adhesive
Coverslip compatibleYesYesYes
Writing surfaceClear endSuperFrost white endSuperFrost blue end
IncludesNA
  • Chamber removal tool
  • Chamber removal tool
 Order nowOrder nowOrder now
Watch Nunc Lab-Tek Chamber Slide System videos
Guide to Using Nunc Lab-Tek Chamber Slide System
Video Player is loading.
Current Time 0:00
Duration 1:59
Loaded: 8.20%
Stream Type LIVE
Remaining Time 1:59
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Guide to Using Nunc Lab-Tek II Chamber Slide System
    Video Player is loading.
    Current Time 0:00
    Duration 2:57
    Loaded: 5.56%
    Stream Type LIVE
    Remaining Time 2:57
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Ordering information
      Compatible cover glass:
      Catalog # Name Size Price (USD) Qty
      171080 Nunc™ Coverglass for 178599 Chamber Slide System, 16 well Case of 96
      71.65
      Online Exclusive
      77.75
      Save 6.10 (8%)
      For Research Use Only. Not for use in diagnostic procedures.