Help prevent contamination with Lab Armor™ Beads
Contaminated cell culture reagents result in distorted data and unpredictable outcomes. Reduce the risk of contamination, achieve reproducible results and have greater confidence in your cell culture by using Gibco cell culture products and Lab Armor Beads.
Lab Armor Beads are dry, metal beads designed to replace water in non-circulating and non-shaking laboratory water baths. Compared to water, Lab Armor Beads are more resistant to bacterial growth and therefore can help prevent the spreading of contamination throughout the lab.
Lab Armor Beads general properties
General properties
  • Metal beads
  • Moisture and gas impermeable
  • High thermal conductivity and retention
  • Smooth & rounded with fluid-like properties
  • Dry, microbial-resistant media
Size and weight
  • Diameter ~ 5-8 mm each
  • Height ~ 1-2 mm each
  • Weight ~ 3.5 lbs per liter
Lab Armor Beads work better than water in your lab bath
A laboratory bath filled with Lab Armor Beads helps to prevent contamination, eliminates tedious maintenance, provides superior performance all while being a more sustainable option.  Read further to learn more about how these features of Lab Armor Beads can benefit you and your lab.
Reduce risk of contamination
  • Eliminates standing water:  Lab Armor Beads reduce the risk of contamination from standing water on your laboratory bench.  Standing water is a breeding ground for bacteria and mold.
  • Promotes sterile work area:  Lab Armor Beads don't drip from bottles like water. You don't need additional precautions with racks, weights or floats. Lab Armor Beads stay clean and organized helping to keep your work area clean and sterile.
Easy use and maintenance
  • Always on:  No need to plan around warm-up times.  You can leave the bath always on to save preparation time. Lab Armor Beads never evaporate!
  • Remains clean and filled:  You can use the time usually spent on emptying, cleaning and filling the water bath for more important tasks. Just spray beads with ethanol every 2–4 weeks.
Sustainable solution
  • Eliminates biocides: liquid disinfectants used for water bath maintenance are harmful to the environment. Periodic use of 70% ethanol is all that is required to keep Lab Armor™ Beads clean and contamination-free.
  • Highly recyclable: formed from ultra-pure, recyclable material.
  • Energy efficient: at 37°C, Lab Armor Beads consume one half of the energy used by a traditional water bath. Set at 65°C, Lab Armor Beads consume less than one quarter of the energy required by water. Download the green fact sheet
Consistent performance
  • Naturally organized: Lab Armor Beads naturally hold items in place, no matter the size or shape; racks, floats and weights are not required.
  • Better temperature stability: Lab Armor Beads have greater temperature uniformity and maintenance than water.
How many liters of beads do I need?
Cross-sectional view of 3 centrifuge tubes embedded in Lab Armor Beads
Enter the maximum fill volume, in L, for your lab bath model below:
You will need to purchase this many liters of beads: 0
Alternatively, you may perform the calculation yourself by following these steps:
  1. Find the maximum fill volume for your lab bath model as specified by the manufacturer.
  2. Multiply the max fill volume by 0.8 to get the volume of Lab Armor Beads needed for your lab bath. (Example: a lab bath with maximum capacity of 10L, would take 8L of Lab Armor Beads. 10L x 0.8 = 8L)
This calculation enables filling the lab bath to 80% of the maximum volume capacity. Please note, Lab Armor Beads may not be suitable for some lab baths with a low fluid detection feature.
Ordering information
Catalog # Name Size Price (USD) Qty
A1254301 Each
635.65
Online Exclusive
670.00
Save 34.35 (5%)
A1254302 Each
1,081.65
Online Exclusive
1,144.00
Save 62.35 (5%)
Optimized temperature control
Thermo Scientific water baths are designed for superior temperature uniformity for
your specific applications and offer unique features such as a seamless stainless steel
interior chamber and epoxy-coated exterior.
Frequently asked questions
GMP manufacturing
Gibco cell culture products are manufactured in facilities compliant with current good manufacturing practices (GMP) and adhere to a robust quality management system, helping to ensure the consistency, reliability, and high quality you can rely on.
Sustainable solutions
We are committed to delivering products that serve the research needs of our customers, while striving to develop them in a way that minimizes our use of natural resources and our impact on the environment.
Resources and support
Resources
Gibco Media Selection Tool
Find the right Gibco media for your application.  Use this interactive tool to select media based upon culture type, components, additives, and format. 
Cell Culture Select Tool
Search your cell line of interest to find product recommendations and resources to help enable a successful cell culture.
Gibco Cell Culture Basics
Learn best practices and fundamentals of cell culture. Topics include aseptic technique and methods for the growth and maintenance of cells in culture.
Cell Culture Videos
Explore this collection of cell culture videos providing valuable insights into techniques, resources, and the latest advancements in the field.
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Support
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support? Contact our expert team for technical and application support of Laboratory Products.
Related products
Lab Armor™ Beads are manufactured by Lab Armor™. Lab Armor™ is a trademark of Lab Armor™, LLC.