Search Thermo Fisher Scientific
Guide RNAs for CRISPR-Cas9 |
When using the CRISPR-Cas9 system to knock out gene expression or knock in a specific mutation, the design, production, and delivery of high-quality guide RNAs (gRNAs) are critical to success. Whether you need transfection-ready gRNAs to use with TrueCut Cas9 Proteins or you need to harness lentivirus to deliver your editing tools to hard-to-transfect or primary cells, we have you covered. Use our gRNA gene search tool to find predesigned gRNAs in either of these formats, or continue to our selection guide below for other choices.
For functional genomics screening, we’ve also packaged our advanced gRNA designs into CRISPR libraries for high-throughput arrayed screening or economical pooled screening.
Select from various CRISPR gRNA formats
Discover quick tools to search and order gRNAs
Learn more about TrueGuide Synthetic gRNAs
Learn more about LentiArray CRISPR gRNAs
Learn more about IVT gRNAs
Innovation is at the core of what we do, and by optimizing our manufacturing procedures we're now able to ship custom and predesigned gRNAs in as little as 5 days to our North American customers.
![]() TrueGuide Synthetic gRNA | ![]() LentiArray Lentiviral gRNA | ![]() Precision gRNA Synthesis Kit | |
Overview |
|
|
|
Format | Synthetic RNA oligo, chemically modified for stability | Lentiviral particles | In vitro-transcribed (IVT) gRNA |
Application | Knockout (predesigned) or knock-in (custom) | Knockout, including library screening | Knockout or knock-in |
Species |
|
| |
Delivery method | Lipid-mediated transfection or electroporation | Lentiviral transduction | Lipid-mediated transfection or electroporation |
Recommended Cas9 format | TrueCut Cas9 Proteins | LentiArray Cas9 Lentivirus | TrueCut Cas9 Proteins |
Controls | Positive and negative controls available | Positive and negative controls available | |
Order now | Order now | Order now |
TrueGuide Synthetic gRNAs are ready-to-transfect CRISPR single guide RNAs (sgRNAs) designed to provide specific and high-efficiency knockout of your target gene. If you need to enrich edited cells, are working with difficult-to-transfect cells, or need long-term sgRNA expression, use our LentiArray CRISPR gRNAs.
Find predesigned TrueGuide gRNAs using our gene search tool
Find predesigned LentiArray CRISPR gRNAs using our selection tool
Have a design of your own? Whether it’s from a journal article, a colleague, or your favorite design tool, use our simple, fast interface to upload the custom gRNA sequences you would like synthesized into a custom TrueGuide gRNA.
Need design assistance for creating an engineered cell line? You can design your CRISPR gene knockout, knock-in, or tagging experiment, and order a complete set of reagents (including gRNAs) for it, with our free TrueDesign Genome Editor software.
To order more than 24 gRNAs for delivery on a 96-well plate, or for gRNAs for use with other nucleases or specific scaffold sequences, please contact us at GEMservices@thermofisher.com.
Invitrogen TrueGuide Synthetic gRNAs are ready-to-transfect CRISPR guide RNAs designed by a proprietary algorithm for high-efficiency editing. These predesigned, guaranteed single guide RNAs offer targeted knockdown with high specificity across the human and mouse genomes.
![]() TrueGuide 1-piece modified Synthetic gRNA | |
Predesigned knockouts for mouse and human cells | |
Custom sequences | |
Validated controls available | |
Available in plate format | |
Ready-to-transfect | |
Chemical modifications* | |
Ideal for primary and stem cells |
*Chemical modifications include 2´O-Methyl analogs and phosphorothioate linkages to increase editing efficiency and protect against nuclease degradation.
gRNA, or guide RNA, is part of the CRISPR-Cas9 system. The gRNA molecule has two components: a target-specific CRISPR RNA (crRNA) and an auxiliary trans-activating crRNA (tracrRNA). The gRNA guides the Cas9 protein to a specific genomic locus via base pairing between the 20-mer crRNA sequence and the target genomic sequence.
sgRNA stands for single guide RNA, as opposed to the conventional, native S. pyogenes CRISPR-Cas9 system that uses a two-part guide in which CRISPR RNA (crRNA) is hybridized with a tracrRNA. An sgRNA combines both RNAs into a single, long RNA. sgRNAs are typically 100 nt in length and can be expressed from a vector or chemically synthesized.
Because the modified sgRNA demonstrates superior efficiency across more cell types than the two-part system, the TrueGuide crRNA:tracrRNA two-part guide RNA product format has been discontinued. This has allowed us to simplify our synthetic gRNA offering and focus on the modified sgRNA.
CRISPR gRNAs are available from Thermo Fisher Scientific as chemically synthesized RNA oligos, packaged lentiviral particles, or generated as in vitro transcribed (IVT) gRNAs using the GeneArt Precision gRNA Synthesis Kit.
Chemical modifications include 2’ O-Methyl analogs and phosphorothioate internucleotide linkages at the 5′ and 3′ ends of the molecule. These modifications enhance editing efficiency by increasing binding to the target site and inhibiting nuclease degradation, respectively.
You can purchase an sgRNA without any chemical modifications by using the custom gRNA order tool.
TrueGuide Synthetic gRNA, when transfected with TrueCut Cas9 Protein, offers maximum editing efficiency across broad cell types while minimizing cell toxicity and innate immune responses. Unlike a plasmid-based system, the RNP system is transient with faster turnover, which minimizes random integrations and off-target effects.
TrueGuide Synthetic gRNA sequences are analyzed by mass spectrometry to verify sequence integrity.
To estimate CRISPR/Cas9 mediated editing efficiency in a pooled cell population, you can use the GeneArt Genomic Cleavage Detection Kit, next-generation sequencing (NGS), or Sanger sequencing. Either sequencing method—NGS of the amplicons from edited populations, or Sanger sequencing of amplicons cloned into plasmids—gives a more accurate estimate of percent editing efficiency and indel types. See the TALEN and CRISPR Validation Tools page for a more extensive discussion of these methods, and the TrueGuide Synthetic gRNA User Guide for protocol guidance.
Invitrogen LentiArray CRISPR gRNA are pre-designed, pre-packaged gRNA lentiviruses designed for efficient gene knockout. LentiArray CRISPR gRNA designs have been created with a proprietary gRNA design algorithm and are optimized for maximum knockout efficiency without compromising specificity.
LentiArray CRISPR gRNAs are available to support pre-screen assay development and post-screen hit validation when performing high-throughput screening experiments with the LentiArray CRISPR libraries. LentiArray CRISPR gRNA should be used with LentiArray Cas9 Lentivirus to perform genome editing experiments in hard-to-transfect cell lines and primary cells.
Figure 3. CRISPR-Cas9-mediated knockout of EGFP with LentiArray gRNA.(A) A431 cells were subjected to EGFR knockout (right column) using LentiArray Cas9 Lentivirus and LentiArray CRISPR gRNA and compared with untreated cells (wild type, left column) and cells treated with Cas9 but no gRNA (Cas9 control, middle column). Cells were stained for EGFR expression (green), nuclei (blue), and cytoskeletal F-actin (red), and analyzed for immunofluorescence. Loss of EGFR expression was observed only in EGFR knockout cells (c, f). (B) Reduction of EGFR signal was also observed via western blot for target protein in EGFR knockout cells vs controls.
The GeneArt Precision gRNA Synthesis Kit is a complete system for rapid synthesis of guide RNA (gRNA).
With this kit, two short single-stranded oligos that code for the target sequence serve as the starting material, and the gRNA template is assembled from these oligos with a T7 promoter in a short “one-pot” PCR reaction. Following this, the assembled product is then used as template in an in vitro transcription (IVT) reaction. A subsequent rapid purification step produces high-yield (>10 µg), high-concentration (>200 ng/µL), transfection-ready gRNA in as little as four hours.
Combining IVT gRNA with our TrueCut Cas9 Protein v2 has been tested in a variety of suspension and adherent cell lines with >70% cleavage efficiencies and no indications of toxicity.
The resulting gRNA can also be co-transfected with our ready-to-transfect CRISPR Nuclease mRNA. Both protein and mRNA Cas9 formats require no plasmid manipulation and are compatible with high-throughput and multiplex genome-wide cell engineering approaches.
For Research Use Only. Not for use in diagnostic procedures.