Front angle on the SimpliAmp Thermal Cycler, including the user interface home screen
The Applied Biosystems SimpliAmp Thermal Cycler is an easy-to use, compact, and accurate thermal cycler designed to fit every lab’s essential PCR workflow. Innovations like a responsive color touch screen and Applied Biosystems VeriFlex temperature control technology enable simple, accurate optimization. And now you can access your SimpliAmp cycler with Thermo Fisher Connect, giving you the freedom to design and securely upload your methods from any mobile device or desktop computer.
Request quote Request demo
Simple, intuitive design for thermocyclers
The SimpliAmp Thermal Cycler has a large color graphical user interface for easy programming and quick checks of your run status at the bench. Simulation modes now make the transition from previous thermal cyclers to the SimpliAmp Thermal Cycler simple, accurate, and efficient.
Screenshot of the SimpliAmp Thermal Cycler splash screen from Applied Biosystems by Thermo Fisher Scientific
Screenshot of the SimpliAmp Thermal Cycler user interface Pre-Heating countdown to 95°C at 1:30:00 with icons such as “Sign In”, “New Method”, “Open Method”, “Incubate”, and “Settings”
Features of SimpliAmp thermal cyclers
  • Intuitive touchscreen interface—Easy-to-use, color touchscreen interface for quick setup and operation
  • VeriFlex Blocks technology—Provides three independent temperature zones for precise PCR optimization
  • Compact design—Space-saving design suitable for any laboratory environment
  • Fast ramp rates—Efficient heating and cooling rates to reduce overall run times
  • Flexible volume range—Accommodates a wide range of reaction volumes from 10 to 100 µL
  • Easy data transfer and monitoring—Has a USB port and compatible with the Thermo Fisher Cloud
  • Pre-programmed protocols—Includes a library of pre-programmed protocols for common applications
  • Energy-efficient—Designed to help minimize power consumption
These features make the SimpliAmp Thermal Cycler a versatile and user-friendly tool for PCR experiments.
Watch SimpliAmp Thermal Cycler customer stories
Watch Quy Nguyen, Research Associate II at Sanford Burnham Institute, discuss her use and recommendation of Applied Biosystems instruments, especially our simple thermal cycler, SimpliAmp Thermal Cycler.
Video Player is loading.
Current Time 0:00
Duration 1:16
Loaded: 0%
Stream Type LIVE
Remaining Time 1:16
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Watch Ji-Young Nho, Medical Laboratory Technologist at U2 BIO, explain her use of Thermo Fisher PCR products and the SimpliAmp Thermal Cycler, an easy-to-use thermal cycler.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Stay in control with Thermal Cycler Fleet Control Software
      Diagram of multiple thermal cyclers connected to one tablet to represent Fleet Control Software
      Manage multiple users, methods, and instruments with a single intuitive interface securely behind your firewall.
      Learn more about Thermal Cycler Fleet Control Software
      Ordering information
      Catalog # Name Size Price (USD) Qty
      A36969 SimpliAmp™ Thermal Cycler Extended Warranty + Plastics Package, REX, 1 package Each
      4,226.51
      Special offer
      Online exclusive
      Ends: 01-Jun-2025
      7,044.19
      Save 2,817.68 (40%)
      A36970 SimpliAmp™ Thermal Cycler Extended Warranty + Plastics Package, ABRC, 1 package Each
      4,284.64
      Special offer
      Online exclusive
      Ends: 01-Jun-2025
      7,141.07
      Save 2,856.43 (40%)
      Catalog # Name Size Price (USD) Qty
      A24811 SimpliAmp™ Thermal Cycler, 1 instrument Each
      4,511.00
      Special offer
      Online exclusive
      Ends: 01-Jun-2025
      6,940.00
      Save 2,429.00 (35%)
      Catalog # Name Size Price (USD) Qty
      A40070 Thermal Cycler Fleet Control Software Each
      2,180.00
      High-performance PCR and qPCR plastics for optimal results
      Five multi-colored PCR tube plates
      Applied Biosystems PCR plastics have been designed and validated to work with our thermal cyclers for more than 25 years. Options for every format and throughput need are available to ensure consistent, precise results for end-point PCR applications. Quickly find the right fit and compatible plastics for your desired model and block formats by using our plastics selection guide.
      Explore online PCR and qPCR plastics selection tool
      Resources
      Thermal Cycler Software and Firmware Files
      Download the latest updates for your instrument
      Interested in private label thermal cyclers?
      Submit an inquiry
      Support
      Service
      We provide a full range of service options for all Applied Biosystems thermal cyclers.
      View service plans
      Multiplex Dynamic Temperature Verification Service
      Instrument qualification service: Have one of our engineers perform temperature qualifications at your site.
      View temperature service options
       Download MDTV flyer
      For Research Use Only. Not for use in diagnostic procedures.