For workflows utilizing in-solution digestion protocols, it is critical to measure protein concentration following sample lysis using a standard protein assay in order to optimize the ratio of sample/protease (w/w) for digestion. It is also necessary to measure protein/peptide concentration upstream of isobaric labeling to ensure that equal amounts of sample are labeled before mixing. Since most protein assays are not sensitive enough to measure peptide concentration after digestion, the easy-to-use Pierce Colorimetric or Fluorescent peptide assays have been designed specifically to improve the sensitivity and reproducibility of quantitation of peptide mixtures.
  • Specific—optimized assays for peptide concentration measurements
  • Optimized—both assays designed for maximum sensitivity, accuracy and reproducibility
  • Convenient—simple protocols with a representative standard included in kit
  • Validated—all products have been fully tested and processed samples have been analyzed using Thermo Scientific™ Mass Spectrometers
Choose the right quantitation assay for your application
 Colorimetric Peptide AssayFluorometric Peptide Assay
Recommended sample typePeptidePeptide
MeasurementColorimetric (480 nm)Fluorescent (Ex. 390, Em. 475 nm)
Linear range15-1000 µg/mL5-1000 µg/mL
Sensitivity15 µg/mL5 µg/mL
Minimum sample volume required20 µL10 µL
Amount detected at minimum sample volume300 ng peptide50 ng peptide
Assay time30 min5 min
Key featureRecommended for TMT-labeled peptidesRecommended for single peptides
 Order nowOrder now
Video Player is loading.
Current Time 0:00
Duration 2:57
Loaded: 5.56%
Stream Type LIVE
Remaining Time 2:57
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Related products
    Catalog # Name Size Price (USD) Qty
    23295 Peptide Digest Assay Standard (1 mg/mL), 1.5 mL Each
    182.65
    Online Exclusive
    204.00
    Save 21.35 (10%)
    The BCA Protein Assay become a popular method for colorimetric detection and quantitation of total protein has a unique advantage over other protein assays because of its compatibility with samples that contain up to 5% surfactants (detergents). Compared to most dye-binding methods, the BCA Assay is affected much less by protein compositional differences, providing greater protein-to-protein uniformity.
    For Research Use Only. Not for use in diagnostic procedures.